Skip to Content
Merck
All Photos(1)

Documents

45462

Supelco

Dithianon

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H4N2O2S2
CAS Number:
Molecular Weight:
296.32
Beilstein:
1325563
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C1C2=C(SC(C#N)=C(S2)C#N)C(=O)c3ccccc13

InChI

1S/C14H4N2O2S2/c15-5-9-10(6-16)20-14-12(18)8-4-2-1-3-7(8)11(17)13(14)19-9/h1-4H

InChI key

PYZSVQVRHDXQSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dithianons belong to the quinone class of fungicides.

Application

Dithianon may be used as an analytical reference standard for the determination of dithianon in:
  • Surface water samples by solid-phase extraction (SPE) and liquid chromatography coupled to quadrupole time-of-flight mass spectrometry (LC-Q-TOFMS).
  • Honeybees by dispersive-SPE followed by liquid and gas chromatography coupled to tandem mass spectrometry (LC-MS/MS and GC-MS/MS) equipped with electrospray ionization (ESI) and multiple reaction monitoring (MRM) detection.
  • Apples by quick, easy, cheap, effective, rugged and safe (QuEChERS) extraction as well as high performance thin-layer chromatography (HPTLC) clean-up procedures and GC-MS/MS.
  • Tomatoes by QuEChERS extraction and LC coupled to triple quadrupole (QqQ) ESI-MS/MS with MRM detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of residues of dithianon in apples by high-performance liquid chromatography.
P G Baker et al.
The Analyst, 109(1), 81-83 (1984-01-01)
Automated online solid-phase extraction-liquid chromatography mass spectrometric analysis of dithianon in water.
Passoni A, et al.
Eur. J. Mass Spectrom., 22(5), 261-267 (2016)
E Sturdík et al.
Chemico-biological interactions, 30(1), 105-104 (1980-04-01)
The inhibition of glycolysis by 2,3-dinitrilo-1,4-dithia-9,10-antraquinone (DDA) in Ehrlich ascites carcinoma (EAC) cells as well as in the investigated respiratory and fermentative strains of yeasts was found to be the result of inactivation of thiol enzymes of this pathway. Increasing
Ivan Halasz et al.
Acta crystallographica. Section B, Structural science, 68(Pt 6), 661-666 (2012-11-21)
The crystal structures of four polymorphs of the pesticide dithianon (5,10-dihydro-5,10-dioxonaphtho[2,3-b]-1,4-dithiine-2,3-dicarbonitrile) have been solved from powder diffraction data and refined using the Rietveld method. Three polymorphs crystallize in non-centrosymmetric space groups. Two polymorphs have Z' > 1. The structures are
Peter Gildemacher et al.
FEMS yeast research, 6(8), 1149-1156 (2006-12-13)
The effect of inoculations with yeasts occurring on apple surfaces and fungicide treatments on the russeting of Elstar apples was studied. Captan, dithianon and a water treatment were implemented to study the interaction between the fungicides, the inoculated yeast species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service