Skip to Content
Merck
All Photos(1)

Key Documents

34234

Supelco

Demeton-S-methyl solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

S-[2-(Ethylthio)ethyl] O,O-dimethyl phosphorothioate, Methyldemetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15O3PS2
CAS Number:
Molecular Weight:
230.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCSCCSP(=O)(OC)OC

InChI

1S/C6H15O3PS2/c1-4-11-5-6-12-10(7,8-2)9-3/h4-6H2,1-3H3

InChI key

WEBQKRLKWNIYKK-UHFFFAOYSA-N

General description

Demeton-S-methyl is a metabolite of the pesticide thiometon, used for food, environmental, and toxicology analyses.

Application

Demeton-S-methyl may be used as an analytical reference standard for the determination of the analyte in rice, wines and baby food products by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Su Jeong et al.
Biochemical and biophysical research communications, 449(3), 263-267 (2014-05-16)
V-type nerve agents, known as VX, are organophosphate (OP) compounds, and show extremely toxic effects on human and animals by causing cholinergic overstimulation of synapses. The bacterial organophosphorus hydrolase (OPH) has attracted much attention for detoxifying V-type agents through hydrolysis
Simo O Pehkonen et al.
Environmental science & technology, 39(8), 2586-2591 (2005-05-12)
The chemical fate of organophosphorus pesticides (OPs) has been proven to depend strongly on the chemistry of their aquatic environment. In particular, metal ions (and metal oxide surfaces) have been known to play an important role in the hydrolytic fate
V Vallet et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(6), 1182-1190 (2007-05-08)
Organophosphorus compounds (OPs), such as pesticides and chemical warfare agents like sarin (GB), soman (GD) and VX, are highly toxic compounds. The OP vapours and their liquid forms are readily absorbed through the skin, therefore, protecting the skin of people
Organophosphorus poisoning at a chemical packaging company.
R D Jones
British journal of industrial medicine, 39(4), 377-381 (1982-11-01)
Sharon Fischer et al.
Archives of biochemistry and biophysics, 434(1), 108-115 (2005-01-05)
Butyrylcholinesterase-encapsulating bioadhesive liposomes are investigated as prophylactic scavengers of organophosphates for local administration to skin, eyes, airways, and lungs-gates through which organophosphates penetrate living systems. The systems were optimized with respect to: encapsulation efficiency; type of bioadhesive ligand bound to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service