Skip to Content
Merck
All Photos(3)

Key Documents

44340

Sigma-Aldrich

AmberChrom 1X8 chloride form

chloride form, strongly basic, 200-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

Quality Level

description

cross-linkage 8%

form

beads

parameter

66 °C OH- form max. temp.

moisture

39-45%

loss

~40% loss on drying, 110 °C

matrix

styrene-divinylbenzene (gel)

particle size

200-400 mesh

operating pH

0-14

capacity

1.2 meq/mL by wetted bed volume

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

AmberChrom 1X8 chloride form is a strongly basic (type I) anion exchange resin for fine chemical processing. AmberChrom has been used to determine trace levels of rhodium and platinum in environmental biological samples.
Strongly basic (type I) anion exchange resin for fine chemical processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Sánchez Rojas et al.
Talanta, 64(1), 230-236 (2008-10-31)
A method for the determination of rhodium in different samples at trace levels is presented. The investigated metal is preconcentrated on a chelating resin microcolumn (1,5-bis(2-pyridyl)-3-sulphophenyl methylene thiocarbonohydrazide (PSTH) immobilized on an anion-exchange resin (Dowex 1x8-200)) placed in the autosampler
M M González García et al.
Analytical and bioanalytical chemistry, 375(8), 1229-1233 (2003-05-07)
A method to determine trace amounts of platinum in different samples based on electrothermal atomic absorption spectrometry is described. The preconcentration step is performed on a chelating resin microcolumn [1,5-bis(2-pyridyl)-3-sulfophenyl methylene thiocarbonohydrazide (PSTH) immobilized on an anion-exchange resin (Dowex 1x8-200)]
P Cañada Rudner et al.
Talanta, 46(5), 1095-1105 (2008-10-31)
1,5-Bis[(2-pyridyl)-3-sulphophenyl methylene] thiocarbonohydrazide (PSTH) immobilized on an anion-exchange resin (Dowex) has been used for the on-line preconcentration of mercury from biological samples and waters prior to its determination by inductively coupled plasma atomic emission spectroscopy. The metal was eluted from
Connor M Blair et al.
Bio-protocol, 10(7), e3581-e3581 (2021-03-05)
Cyclic nucleotide degrading phosphodiesterase (PDE) enzymes are crucial to the fine tuning of cAMP signaling responses, playing a pivotal role in regulating the temporal and spatial characteristics of discrete cAMP nanodomains and hence the activity of cAMP-effector proteins. As a
Daniel Dar et al.
Science (New York, N.Y.), 373(6556) (2021-08-14)
Capturing the heterogeneous phenotypes of microbial populations at relevant spatiotemporal scales is highly challenging. Here, we present par-seqFISH (parallel sequential fluorescence in situ hybridization), a transcriptome-imaging approach that records gene expression and spatial context within microscale assemblies at a single-cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service