Skip to Content
Merck
All Photos(1)

Documents

43318

Supelco

Citral

analytical standard

Synonym(s):

3,7-Dimethyl-2,6-octadienal, Geranial and neral mixture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
Beilstein:
1721871
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5 (vs air)

vapor pressure

0.2 mmHg ( 200 °C)

Assay

≥95.0% (cis + trans, GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.487-1.489
n20/D 1.488 (lit.)

bp

229 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

suitability

conforms to structure for Proton NMR spectrum

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C=C(C)CC\C=C(\C)C

InChI

1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3

InChI key

WTEVQBCEXWBHNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citral usually is a key component of lemon scented essential oils and also in volatile oils of plants like ginger, which can exist like a mixture of two isomers, cis-isomer neral and trans-isomer geranial. It can serve as potential candidate used in the treatment of tumours. It can mainly find applications as a food additive and as a fragrance in cosmetics.
Citral, a volatile compound, is considered as an important raw material for lemon scented tea tree oil and Cymbopogon citratus volatile oil. It is used as an aroma ingredient in food products, aromatherapy, cosmetic industry, scenting of soaps and toiletries. The antimicrobial activity of this compound on the airborne microbes is investigated.

Application

Citral may be used as an analytical reference standard for the quantification of the analyte in:
  • Essential oils using reversed-phase high performance liquid chromatography with photodiode array detection (RP-HPLC-PDA), further confirmed by gas-chromatography coupled to mass spectrometry (GC-MS).
  • Cymbopogon citratus volatile oil using high performance liquid chromatography with UV detection (HPLC-UV) and gas chromatography coupled to mass spectrometry (GC-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - DIN 51758

Flash Point(C)

98 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Citral Assay, Determination of Citral in Lemongrass and Citrus Oils by Condensation with Barbituric Acid
Levi L and Laughton MP
Journal of Agricultural and Food Chemistry, 7(12), 850-859 (1959)
Citral is a new inducer of caspase-3 in tumor cell lines
Dudai N, et al.
Planta Medica, 71(05), 484-488 (2005)
Antipyretic Effects of Citral and Possible Mechanisms of Action
Emilio-Silva.TM, et al.
Inflammation, 1-7 (2017)
Antimicrobial effect of trans-cinnamaldehyde,(?)-perillaldehyde,(?)-citronellal, citral, eugenol and carvacrol on airborne microbes using an airwasher
Sato K, et al.
Biological & Pharmaceutical Bulletin, 29(11), 2292-2294 (2006)
Development and validation of reverse phase high performance liquid chromatography for citral analysis from essential oils
Gaonkar R, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1036, 50-56 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service