Skip to Content
Merck
All Photos(1)

Key Documents

43016

Supelco

Ginsenoside Rg5

analytical standard

Synonym(s):

(3β, 12β,20E)-12-Hydroxydammara-20(22),24-dien-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O12
CAS Number:
Molecular Weight:
767.00
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O)CO)O[C@@H]3C([C@H]4[C@@]([C@@H]5[C@]([C@]6([C@H]([C@@H](C5)O)[C@H](CC6)\C(=C\CC=C(C)C)\C)C)(CC4)C)(CC3)C)(C)C

InChI

1S/C42H70O12/c1-21(2)10-9-11-22(3)23-12-16-42(8)30(23)24(45)18-28-40(6)15-14-29(39(4,5)27(40)13-17-41(28,42)7)53-38-36(34(49)32(47)26(20-44)52-38)54-37-35(50)33(48)31(46)25(19-43)51-37/h10-11,23-38,43-50H,9,12-20H2,1-8H3/b22-11+/t23-,24-,25-,26-,27+,28-,2

InChI key

NJUXRKMKOFXMRX-RNCAKNGISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginsenoside Rg5 is a diol-containing ginsenoside, that occurs naturally in Panax ginseng. It is found to be a promising agent in the treatment of Alzheimer′s disease.

Application

Ginsenoside Rg5 may be used as a reference standard in the determination of ginsenoside Rg5 in Korean red ginseng using high performance liquid chromatography coupled with evaporative light scattering detection (HPLC-ELSD).

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginsenoside-Rg5 suppresses cyclin E-dependent protein kinase activity via up-regulating p21Cip/WAF1 and down-regulating cyclin E in SK-HEP-1 cells.
K Y L, et al.
Anticancer Research, 17(2A), 1067-1072 (1997)
Ginsenoside Rg5 improves cognitive dysfunction and beta-amyloid deposition in STZ-induced memory impaired rats via attenuating neuroinflammatory responses.
Chu S, et al.
International Journal of Immunopharmacology, 19(2), 317-326 (2014)
Simultaneous quantification of 14 ginsenosides in Panax ginseng CA Meyer (Korean red ginseng) by HPLC-ELSD and its application to quality control.
Kim NS, et al.
Journal of Pharmaceutical and Biomedical Analysis, 45(1), 164-170 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service