Skip to Content
Merck
All Photos(2)

Key Documents

39961

Supelco

Cannabidiolic acid

analytical standard

Synonym(s):

CBDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule I; Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

suitability

in accordance for H-NMR

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CCCCCc1cc(O)c([C@@H]2C=C(C)CC[C@H]2C(C)=C)c(O)c1C(O)=O

InChI

1S/C22H30O4/c1-5-6-7-8-15-12-18(23)20(21(24)19(15)22(25)26)17-11-14(4)9-10-16(17)13(2)3/h11-12,16-17,23-24H,2,5-10H2,1,3-4H3,(H,25,26)/t16-,17+/m0/s1

InChI key

WVOLTBSCXRRQFR-DLBZAZTESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cannabidiolic acid belongs to the group of cannabinoids, a class of psychoactive and non-psychoactive compounds produced in the Cannabis (marijuana) plant. It is an acidic precursor,(4) which shows potential therapeutic efficacy in the treatment of pain, mood disorders, and inflammatory diseases.[1][2][3]

Application

Cannabidiolic acid has been used as an analytical standard for the quantification of the analyte in biomass, extracts of different varieties of cannabis,[1] and commercial cannabis products[2] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roger G Pertwee et al.
British journal of pharmacology, 175(1), 100-112 (2017-10-24)
The aim of this study was to compare the abilities of cannabidiolic acid methyl ester (HU-580) and cannabidiolic acid (CBDA) to enhance 5-HT We investigated the effects of HU-580 and CBDA on (i) activation by 8-hydroxy-2-(di-n-propylamino)tetralin of human 5-HT HU-580
Szabolcs Fekete et al.
Journal of pharmaceutical and biomedical analysis, 155, 116-124 (2018-04-09)
A generic liquid chromatographic method development workflow was developed and successfully applied to the analysis of phytocannabinoids and Cannabis sativa extracts. Our method development procedure consists in four steps: At the end, this method development workflow was performed in less
Bryan Duffy et al.
Toxics, 8(1) (2020-01-30)
Beginning in June of 2019, there was a marked increase in reported cases of serious pulmonary injury associated with vaping. The condition, referred to as e-cigarette or vaping product use-associated lung injury (EVALI), does not appear to involve an infectious
Kazuhito Watanabe et al.
Toxicology, 206(3), 471-478 (2004-12-14)
The progesterone 17alpha-hydroxylase activity, which is one of the steroidogenic enzymes in rat testis microsomes, was significantly inhibited by crude marijuana extracts from Delta(9)-tetrahydrocannabinolic acid (THCA)- and cannabidiolic acid (CBDA)-strains. Delta(9)-Tetrahydrocannabinol, cannabidiol and cannabinol also inhibited the enzymatic activity with
Cannabidiolic?acid synthase, the chemotype?determining enzyme in the fiber?type Cannabis sativa
Taura F, et al.
Febs Letters, 581, 2929-2934 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service