Skip to Content
Merck
All Photos(2)

Documents

36538

Supelco

Triclopyr 2-butoxyethylester

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16Cl3NO4
CAS Number:
Molecular Weight:
356.63
Beilstein:
8155305
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCOCCOC(=O)COc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C13H16Cl3NO4/c1-2-3-4-19-5-6-20-11(18)8-21-13-10(15)7-9(14)12(16)17-13/h7H,2-6,8H2,1H3

InChI key

IVDRCZNHVGQBHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Johansen et al.
Archives of environmental contamination and toxicology, 19(4), 610-616 (1990-07-01)
The toxicity of Garlon4, the ethylene glycol butyl ether ester formulation of the herbicide tryclopyr, to juvenile coho salmon (Oncorhynchus kisutch) was investigated at several lethal and sublethal concentrations. Fish behavior, random activity and oxygen uptake were monitored. Coho salmon
Triclopyr persistence in northern Idaho forest vegetation.
S G Whisenant et al.
Bulletin of environmental contamination and toxicology, 42(5), 660-665 (1989-05-01)
Acute toxicity to juvenile Pacific salmonids of Garlon 3A, Garlon 4, triclopyr, triclopyr ester, and their transformation products: 3,5,6-trichloro-2-pyridinol and 2-methoxy-3,5,6-trichloropyridine.
M T Wan et al.
Bulletin of environmental contamination and toxicology, 39(4), 721-728 (1987-10-01)
Acute toxicity of Garlon 4 and Roundup herbicides to salmon, Daphnia, and trout.
J A Servizi et al.
Bulletin of environmental contamination and toxicology, 39(1), 15-22 (1987-07-01)
Christopher A Nowak et al.
Environmental management, 36(2), 237-247 (2005-07-05)
There are a wide variety of different herbicide treatment methods used to remove single trees. Each method (cut-stump, basal, foliar) has a unique amount of off-target disturbance that should be considered in selecting a treatment for use in management. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service