Skip to Content
Merck
All Photos(1)

Documents

36780

Supelco

Fluroxypyr-1-methylheptyl ester

PESTANAL®, analytical standard

Synonym(s):

1-Methylheptyl (4-amino-3,5-dichloro-6-fluoro-2-pyridyloxy)acetate, Fluroxypyr meptyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21Cl2FN2O3
CAS Number:
Molecular Weight:
367.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCCCC(C)OC(=O)COc1nc(F)c(Cl)c(N)c1Cl

InChI

1S/C15H21Cl2FN2O3/c1-3-4-5-6-7-9(2)23-10(21)8-22-15-12(17)13(19)11(16)14(18)20-15/h9H,3-8H2,1-2H3,(H2,19,20)

InChI key

OLZQTUCTGLHFTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluroxypyr-1-methylheptyl ester is a selective post-emergent and systemic herbicide.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Residue Analysis of Fluroxypyr-meptyl in Wheat and Soil by GC?ECD
Hu J-Y, et al.
Chromatographia, 74, 291-296 (2011)
Christina E Cowan-Ellsberry et al.
Chemosphere, 70(10), 1804-1817 (2007-10-02)
National and international chemical management programs are assessing thousands of chemicals for their persistence, bioaccumulative and environmental toxic properties; however, data for evaluating the bioaccumulation potential for fish are limited. Computer based models that account for the uptake and elimination
Effects of different dilution water types on the acute toxicity to juvenile Pacific salmon and rainbow trout of fluroxypyr formulated product XRM-5084.
M T Wan et al.
Bulletin of environmental contamination and toxicology, 49(6), 914-921 (1992-12-01)
P G Hewitt et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(7), 748-754 (2000-06-20)
Percutaneous absorption of pesticides is a major determinant for risk assessment. Furthermore, cutaneous metabolism plays a role in penetration of certain chemicals. Therefore, the aim of these studies was to determine the transdermal metabolism of three related compounds [the herbicide
P G Hewitt et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(7), 755-759 (2000-06-20)
Fluroxypyr methyl ester (FPM) and the herbicide fluroxypyr methylheptyl ester (FPMH) are completely hydrolyzed during penetration through human and rat skin in vitro to the acid metabolite, fluroxypyr (FP) (). This article presents additional studies to determine the enzyme kinetics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service