Skip to Content
Merck
All Photos(1)

Key Documents

36170

Supelco

Mirex

PESTANAL®, analytical standard

Synonym(s):

Perchloropentacyclodecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10Cl12
CAS Number:
Molecular Weight:
545.54
Beilstein:
5605269
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC1(Cl)C2(Cl)C3(Cl)C4(Cl)C(Cl)(Cl)C5(Cl)C3(Cl)C1(Cl)C5(Cl)C24Cl

InChI

1S/C10Cl12/c11-1-2(12)7(17)4(14)3(13,5(1,15)9(7,19)20)6(1,16)10(21,22)8(2,4)18

InChI key

GVYLCNUFSHDAAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mirex is an organochlorine insecticide used for control of the imported fire ant (Solenopsis richteri Forel and Solenopsis invicta Buren).

Application

Mirex may be used as an internal reference standard to evaluate instrument performance during the determination of polybrominated diphenyl ethers and polychlorinated biphenyls in human serum and plasma by gas chromatography–tandem mass spectrometry method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirex residues in human adipose tissue.
Kutz F W, et al.
Environmental Entomology, 3(5), 882-884 (1974)
Simultaneous determination of polybrominated diphenyl ethers and polychlorinated biphenyls by gas chromatography?tandem mass spectrometry in human serum and plasma.
Lin YP, et al.
Talanta, 113, 41-48 (2013)
Mirex. An overview.
E M Waters et al.
Environmental research, 14(2), 212-222 (1977-10-01)
Lawrence M Schell et al.
Physiology & behavior, 99(2), 246-253 (2009-10-06)
Endocrine disruption has become a significant human health concern, but is difficult to study outside of the laboratory for several reasons including the multiplicity of exposures, the difficulty in assessing each exposure, and the variety of possible outcomes among human
Juliana Leonel et al.
Marine pollution bulletin, 60(3), 412-418 (2009-11-26)
Selected POPs were analyzed in blubber samples of Pontoporia blainvillei from Southern Brazil to appraise temporal trend over a 10 year period (1994-2004). Overall, levels of POPs were relatively low, especially when compared to Northern Hemisphere concentrations. Apart from Mirex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service