Skip to Content
Merck
All Photos(1)

Key Documents

48103

Supelco

Toxaphene solution

certified reference material, 1000 μg/mL in isooctane

Synonym(s):

Camphechlor solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8081

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

ClC21C(C(C(C2Cl)Cl)(C(=C)C1(CCl)CCl)Cl)(Cl)Cl

InChI

1S/C10H8Cl8/c1-4-7(2-11,3-12)9(16)6(14)5(13)8(4,15)10(9,17)18/h5-6H,1-3H2

InChI key

OEJNXTAZZBRGDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaoqiong Tian et al.
Se pu = Chinese journal of chromatography, 30(1), 14-20 (2012-06-07)
A novel method for the determination of representative toxaphene congeners in traditional Chinese herbal medicines was developed. Ginseng and Milkvetch Root were selected as the samples and seven toxaphene congeners were selected as the monitoring objects. The samples were extracted
Seonock Woo et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 156(1), 37-41 (2012-04-14)
We evaluated toxaphene-induced acute toxicity in Hydra magnipapillata. The median lethal concentrations of the animals (LC(50)) were determined to be 34.5 mg/L, 25.0 mg/L and 12.0 mg/L after exposure to toxaphene for 24 h, 48 h and 72 h, respectively.
Thomas Kapp et al.
Chemosphere, 82(1), 32-36 (2010-11-13)
For decades, toxaphene had been used as a major chloropesticide. Degradation of the multicomponent mixture in the environment was mainly reported to be due to anaerobic dechlorination and hydrodechlorination. Little was known about oxidative transformation processes and the potential hydroxylated
Juan José Alava et al.
Environmental toxicology and chemistry, 30(7), 1677-1688 (2011-04-22)
Persistent organic pollutants (POPs) are recognized manmade threats to sea turtle populations, but substantial uncertainty exists surrounding their exposure to contaminants and their sensitivity to toxic effects. This uncertainty creates difficulty for conservation managers to make informed decisions for the
Wenjian Lao et al.
Journal of chromatography. A, 1270, 262-268 (2012-11-20)
Determinative methods based on gas chromatography-negative chemical ionization mass spectrometry (GC-NCI/MS) provide improved sensitivity and specificity for toxaphene in environmental samples, but are subject to misidentification due to oxygen reaction in the presence of polychlorinated biphenyls (PCBs). The goal of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service