Skip to Content
Merck
All Photos(1)

Documents

339075

Supelco

Boric anhydride

99.98% trace metals basis

Synonym(s):

Boron trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2O3
CAS Number:
Molecular Weight:
69.62
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38120103
PubChem Substance ID:
NACRES:
NA.21

grade

for inorganic trace analysis

Quality Level

vapor density

>1 (vs air)

Assay

99.98% trace metals basis

form

powder

mp

450 °C (lit.)

density

2.46 g/mL at 25 °C (lit.)

SMILES string

O=BOB=O

InChI

1S/B2O3/c3-1-5-2-4

InChI key

JKWMSGQKBLHBQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boric anhydride in sulfuric acid may be used as a scrubber to remove the hydrogen fluoride gas formed as a byproduct during the selective reduction of ketones to hydrocarbons using triethylsilane and boron trifluoride. In combination with hydroxyapatite, it may be used to catalyze the vapor-phase Beckmann rearrangement process to form nylon-6.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vapor-phase Beckmann rearrangement of cyclohexanone oxime over boria-hydroxyapatite catalyst
Izumi Y, et al.
Chemistry Letters (Jpn), 12(10), 1649-1652 (1983)
Reduction of ketones to hydrocarbons with triethylsilane: m-nitroethylbenzene
Fry JL, et al.
Organic Syntheses, 60, 108-108 (1981)
Minh Tho Nguyen et al.
The journal of physical chemistry. A, 113(17), 4895-4909 (2009-04-01)
Thermochemical properties of a set of small boron (B(n)) and boron oxide (B(n)O(m)) clusters, with n = 1-4 and m = 0-3, their anions, and the B(4)(2-) dianion, were calculated by using coupled-cluster theory CCSD(T) calculations with the aug-cc-pVnZ (n
Xinyong Tao et al.
Nano letters, 8(2), 505-510 (2008-01-09)
Mg2B2O5 nanowires with (010) twins were synthesized for the first time using a catalyst-free method. The microstructure of the Mg2B2O5 nanowires has been extensively studied by cross-sectional high-resolution transmission electron microscopy. Nanoindentation tests were performed directly on individual nanowires to
S O Hruszkewycz et al.
Physical review letters, 109(18), 185502-185502 (2012-12-12)
The availability of ultrafast pulses of coherent hard x rays from the Linac Coherent Light Source opens new opportunities for studies of atomic-scale dynamics in amorphous materials. Here, we show that single ultrafast coherent x-ray pulses can be used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service