Merck
  • Catalyst-free synthesis, structural, and mechanical characterization of twinned Mg2B2O5 nanowires.
Catalyst-free synthesis, structural, and mechanical characterization of twinned Mg2B2O5 nanowires.
Nano letters (2008-01-09)
Xinyong Tao, Xiaodong Li
ABSTRACT
Mg2B2O5 nanowires with (010) twins were synthesized for the first time using a catalyst-free method. The microstructure of the Mg2B2O5 nanowires has been extensively studied by cross-sectional high-resolution transmission electron microscopy. Nanoindentation tests were performed directly on individual nanowires to probe their mechanical properties. It was found that the twinned Mg2B2O5 nanowires achieve comparable hardness but 19% decrease in elastic modulus compared to their bulk counterpart. The elastic softening mechanisms of the Mg2B2O5 nanowires are discussed with reference to their twin defects, size, and surface effects.
Sigma-Aldrich
Boric anhydride, puriss. p.a., ≥98% (T)
Sigma-Aldrich
Boric anhydride, 99.999% trace metals basis
Supelco
Boric anhydride, 99.98% trace metals basis
Sigma-Aldrich
Boric anhydride, granulated, ≥98.0% (T)