Skip to Content
Merck
All Photos(1)

Key Documents

33528

Sigma-Aldrich

4-Aminoantipyrine

puriss. p.a., reag. Ph. Eur., ≥99%

Synonym(s):

4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Beilstein:
181635
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Agency

USP/NF
reag. Ph. Eur.

Quality Level

grade

puriss. p.a.

Assay

≥99%

ign. residue

≤0.1%

loss

≤0.5% loss on drying

mp

105-110 °C (lit.)
107-109 °C

SMILES string

CN1N(c2ccccc2)C(=O)C(N)=C1C

InChI

1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3

InChI key

RLFWWDJHLFCNIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminoantipyrine forms heterocyclic Schiff bases, by reaction with various aldehydes and oximes. These Schiff bases form stable complexes with transition metals.[1]

Application

  • Phenol impurity and wastewater analysis: Research demonstrated the application of 4-Aminoantipyrine in spectrophotometric determination of phenol impurities in phenoxyethanol and for assessing phenol index in drinking water and municipal wastewater, utilizing salting-out assisted liquid phase microextraction (Roustaei et al., 2024).
  • Advanced sensor development for tea polyphenols: Utilization of 4-Aminoantipyrine in developing a novel hybrid sensor array, integrated with polyphenol oxidase and its nanozymes, aided by machine learning to identify tea polyphenols and Chinese teas, showcasing its role in innovative sensor technology (Yang et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Stange et al.
Scientific reports, 10(1), 6149-6149 (2020-04-11)
Low birth weight (LBW) can cause lifelong impairments in muscle development and growth. Satellite cells (SC) and their progeny are crucial contributors to myogenic processes. This study provides new data on LBW in piglets combining insights on energy metabolism, muscle
Transition metal complexes with Schiff-base ligands: 4-aminoantipyrine based derivatives-a review.
Raman N, et al.
Journal of Coordination Chemistry, 62(5, 691-709 (2009)
Synthesis, structural determination and antibacterial activity of compounds derived from vanillin and 4-aminoantipyrine.
Vaghasiya YK, et al.
J. Serb. Chem. Soc., 69(12), 991-998 (2004)
Tao Sun et al.
Frontiers in bioengineering and biotechnology, 8, 237-237 (2020-04-09)
Hypoxia not only alters tumor microenvironment but leads to the tumor progression and metastasis as well as drug resistance. As a promising strategy, photodynamic therapy (PDT) can inhibit tumor by catalyzing O2 to cytotoxic reactive oxygen species. However, its effects
Lucas de Melo da Silva et al.
The Science of the total environment, 631-632, 1079-1088 (2018-05-08)
Electro-oxidation with electrogenerated H2O2 (EO-H2O2) was applied to treat acidic aqueous solutions of 4-aminoantipyrine (4-AA), a persistent drug metabolite of dipyrone, in sulfate medium. Trials were made using a boron-doped diamond anode in the presence of H2O2 electrogenerated on site.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service