Skip to Content
Merck
All Photos(1)

Key Documents

32425

Supelco

Cyclosporin A

VETRANAL®, analytical standard

Synonym(s):

Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Gene Information

human ... PPIA(5478)

Looking for similar products? Visit Product Comparison Guide

General description

Cyclosporin A (CsA) is a fungal metabolite, a cyclic undecapeptide. It has highly specific immunosuppressive effects with low myelotoxicity, thereby making it a valuable drug in human organ transplantation. Its immunosuppressive activity is based upon its ability to inhibit early steps of T-lymphocyte activation such as lymphokine gene transcription.
Cyclosporin A has been used to:
  • Prevent rejection of kidney and liver transplants.
  • In graft versus host disease.
  • Treatment of autoimmune diseases.
  • Treatment of Schistosomiasis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.
Potent immunosuppressant; inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα; blocks cytochrome c release from mitochondria.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclosporin A specifically inhibits function of nuclear proteins involved in T cell activation.
Emmel, Elizabeth A., et al.
Science, 246 (4937), 1617-1617 (1989)
Cyclosporin A inhibits T-cell growth factor gene expression at the level of mRNA transcription.
Kronke, M., et al.
Proceedings of the National Academy of Sciences of the USA, 81 (16), 5214-5218 (1984)
R E Handschumacher et al.
Science (New York, N.Y.), 226(4674), 544-547 (1984-11-02)
Cyclophilin, a specific cytosolic binding protein responsible for the concentration of the immunosuppressant cyclosporin A by lymphoid cells, was purified to homogeneity from bovine thymocytes. Cation-exchange high-performance liquid chromatography resolved a major and minor cyclophilin species that bind cyclosporin A
Xiao-Ling Liu et al.
Bioorganic & medicinal chemistry, 16(1), 171-180 (2007-10-30)
A library of chalcones with basic functionalities were screened for inhibition of P-glycoprotein (Pgp, ABCB1) by the calcein-AM accumulation assay on MDCKII/MDR1 cells. Three members that had ring A substituted with 5-(1-ethylpiperidin-4-yl) and 2,4-dimethoxy groups were found to increase calcein-AM
Mi-Yeon Jang et al.
Journal of medicinal chemistry, 54(2), 655-668 (2010-12-22)
Herein we describe the synthesis and in vitro and in vivo activity of thiazolo[5,4-d]pyrimidines as a novel class of immunosuppressive agents, useful for preventing graft rejection after organ transplantation. This research resulted in the discovery of a series of compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service