Skip to Content
Merck

Skip To

C-093

Cyclosporin A solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O=C(N(C)[C@@H](CC(C)C)C(N[C@@H](C(C)C)C(N(C)[C@H](C(N[C@@H](C)C(N[C@H](C)C(N(C)[C@@H](CC(C)C)C(N(C)[C@@H](CC(C)C)C(N([C@H](C(C)C)C(N(C)[C@@H]([C@@H]([C@H](C)C/C=C/C)O)C1=O)=O)C)=O)=O)=O)=O)=O)CC(C)C)=O)=O)CN(C)C([C@@H](N1)CC)=O

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50?,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-VPYFBNRVSA-N

General description

A Certified Spiking Solution® of an immunosuppressant and narrow therapeutic index drug used to prevent rejection in organ transplantation. This certified calibration material is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the drug′s narrow therapeutic range.  The Snap-N-Spike® format allows laboratories to eliminate weighing operations of hazardous substances such as cyclosporin A, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML101832425PHR1092
description

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

description

1 mg/mL in DMSO

description

VETRANAL®, analytical standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

single component solution

format

-

format

neat

format

neat

form

liquid

form

DMSO solution

form

-

form

-

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

300

grade

certified reference material

grade

-

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

storage temp.

−70°C

storage temp.

−20°C

storage temp.

-

storage temp.

-10 to -25°C


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters