Skip to Content
Merck

Skip To

A9231

Acridine Orange solution

2% in H2O

Synonym(s):

3,6-Acridinediamine, N,N,N′,N′-tetramethyl-, monohydrochloride, 3,6-Bis(dimethylamino)acridine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H19N3 · HCl
CAS Number:
Molecular Weight:
301.81
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

shelf life

18 mo.

concentration

2% in H2O

technique(s)

microbe id | staining: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

N(C)(C)c1cc2nc3c(cc2cc1)ccc(c3)N(C)C

InChI

1S/C17H19N3/c1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14/h5-11H,1-4H3

InChI key

DPKHZNPWBDQZCN-UHFFFAOYSA-N

General description

Acridine Orange (C.l. 46005) is an acridine dye with a ring structure and overall small planar conjugated system.

Application

Acridine orange is a DNA intercalating dye that is a recommended DNA stain for flow cytometry. It differentially stains double-stranded and single-stranded nucleic acids.

It also has the following applications-
  • staining of RNA and acid mucins
  • detecting apoptosis
  • staining blood smears to diagnose malaria
  • identifying and enumerating granulocytes in fish
  • staining eukaryotic cells to demonstrate low pH structures

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8097235474A6014
technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

nucleic acid detection: suitable

concentration

2% in H2O

concentration

10 mg/mL in H2O

concentration

-

concentration

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

-

form

liquid

form

solution

form

powder

form

powder

shelf life

18 mo.

shelf life

-

shelf life

-

shelf life

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Matthew P Harris et al.
PLoS genetics, 4(10), e1000206-e1000206 (2008-10-04)
The genetic basis of the development and variation of adult form of vertebrates is not well understood. To address this problem, we performed a mutant screen to identify genes essential for the formation of adult skeletal structures of the zebrafish.
Hong-xiang Wang et al.
Zhonghua yi xue za zhi, 92(26), 1828-1831 (2012-09-05)
To ascertain the epididymal sperm nuclear maturity in obstructive azoospermia (OA) patients. A total of 81 infertile males from andriatry clinic of Renji hospital were selected, including 32 OA patients (OA group) and 49 asthenospermia patients (asthenospermia group). Another 32
Grace Emily Okuthe
Acta histochemica, 115(2), 178-184 (2012-07-17)
Oogenesis involves a sequence of cellular divisions and developmental changes leading to the formation of oocytes, whose role in development is to transfer genomic information to the next generation. During this process, the gene expression pattern changes considerably concomitant with



Global Trade Item Number

SKUGTIN
A9231-10ML04061838075659

Questions

Reviews

No rating value

Active Filters