Skip to Content
Merck
All Photos(2)

Key Documents

31611

Supelco

Phenthoate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17O4PS2
CAS Number:
Molecular Weight:
320.36
Beilstein:
2474108
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)C(SP(=S)(OC)OC)c1ccccc1

InChI

1S/C12H17O4PS2/c1-4-16-12(13)11(10-8-6-5-7-9-10)19-17(18,14-2)15-3/h5-9,11H,4H2,1-3H3

InChI key

XAMUDJHXFNRLCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Imamura et al.
Toxicology and applied pharmacology, 70(1), 140-147 (1983-08-01)
Malathion and phenthoate carboxylesterase activities were investigated in pulmonary alveolar macrophages (PAM) in Sprague-Dawley rats. PAM was found to be capable of hydrolyzing phenthoate at a faster rate than malathion. Oral administration to rats with O,O,S-trimethyl phosphorothioate (OOS-Me), a pneumotoxic
Usefulness of gas chromatography/negative ion chemical ionization mass spectrometry for detection of an organophosphate pesticide in a victim.
H Hattori et al.
Medicine, science, and the law, 26(4), 263-269 (1986-10-01)
S Banerjee et al.
Ecotoxicology and environmental safety, 29(3), 265-275 (1994-12-01)
Histopathological changes in the head and trunk kidneys of Channa punctatus induced by chronic nonlethal levels of Elsan (211 ppb), mercuric chloride (16.7 ppb), and aqueous ammonia (15.64 ppm) were studied on 7, 28, 63, and 90 days of exposure.
S Banerjee et al.
Ecotoxicology and environmental safety, 31(1), 62-68 (1995-06-01)
Histopathological changes in the intestine of Channa punctatus induced by chronic nonlethal levels of Elsan (211 ppb), mercury (16.7 ppb), and ammonia (15.64 ppm) were studied at 7-day intervals for 90 days and the data were presented only for days
K S Rao et al.
Ecotoxicology and environmental safety, 9(1), 107-111 (1985-02-01)
The levels of total lipids, free fatty acids, cholesterol, and lipase activity were studied in selected tissues of Channa punctatus Bloch during individual and combined exposures of carbaryl and phenthoate. The total lipid levels decreased with elevated levels of free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service