Skip to Content
Merck
All Photos(3)

Key Documents

293105

Sigma-Aldrich

Tetrahydropyran

anhydrous, 99%

Synonym(s):

Pentamethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102436
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
88 °C (lit.)

grade

anhydrous

Quality Level

vapor density

3 (vs air)

Assay

99%

form

liquid

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.42 (lit.)

bp

88 °C (lit.)

mp

−45 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

SMILES string

C1CCOCC1

InChI

1S/C5H10O/c1-2-4-6-5-3-1/h1-5H2

InChI key

DHXVGJBLRPWPCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydropyran (THP) is a saturated cyclic ether. It is commonly used as a solvent in organic synthesis because of its strong hydrophobicity and low vaporization temperature, which enable easy recovery and reuse of the solvent. This makes the entire synthetic process more environmentally friendly.

Application

Tetrahydropyran can be used as a solvent in various classical organic reactions such as radical reactions, palladium-catalyzed coupling, and Grignard reactions.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baoling Lai et al.
Developmental neurobiology, 78(9), 859-872 (2018-07-20)
The cyclic nucleotide cGMP is an intracellular second messenger with important roles in neuronal functions and animals' behaviors. The phosphodiesterases (PDEs) are a family of enzymes that hydrolyze the second messengers cGMP and cAMP. Inhibition of phosphodiesterase 9 (PDE9), a
Zhaotian Zhang et al.
International ophthalmology, 39(7), 1543-1551 (2018-06-24)
To introduce novel and easy techniques for 27-gauge silicone oil (SO) infusion and removal. Consecutive patients treated with 27-gauge pars plana vitrectomy (PPV) plus SO infusion (Infusion Group) and scheduled to have SO removal (Removal Group) were prospectively included, respectively.
Flame chemistry of tetrahydropyran as a model heteroatomic biofuel.
Labbe NJ, et al.
Proceedings of the Combustion Institute, 34(3), 259-267 (2013)
Phase behaviour of gas hydrates of carbon dioxide in the presence of tetrahydropyran, cyclobutanone, cyclohexane and methylcyclohexane.
Mooijer-Van DH, et al.
Fluid Phase Equilibria, 182(1), 97-110 (2001)
Angélica Quintero-Flórez et al.
Journal of agricultural and food chemistry, 65(36), 7900-7907 (2017-08-15)
Volatile compounds are responsible for some sensory characteristics of virgin olive oil (VOO); however, they have not been studied from a nutritional point view. In this work, the effect of the simulated digestion on VOO volatile compounds responsible for green

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service