Skip to Content
Merck
All Photos(3)

Documents

140732

Sigma-Aldrich

N,N-Dimethylformamide dimethyl acetal

technical grade, 94%

Synonym(s):

1,1-Dimethoxy-N,N-dimethylmethylamine, 1,1-Dimethoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH(OCH3)2
CAS Number:
Molecular Weight:
119.16
Beilstein:
506020
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

94%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.396 (lit.)

bp

102-103 °C/720 mmHg (lit.)

density

0.897 g/mL at 25 °C (lit.)

SMILES string

COC(OC)N(C)C

InChI

1S/C5H13NO2/c1-6(2)5(7-3)8-4/h5H,1-4H3

InChI key

ZSXGLVDWWRXATF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylformamide dimethyl acetal (DMF-DMA) can be used as: 
  • An organocatalyst for the synthesis of 1,2-disubstituted-3,4-dihydronaphthalenes by the reaction of vinylarenes and electron-deficient alkynes.       
  • A building block for the preparation of heterocyclic compounds such as enamines and amidines via formylation of methylene and amino groups.       
  • A reagent in the synthesis of 1,4-diaryl-piperazine-2,5-diones by reacting with chloroacetamides.        
  • A reagent along with enamine and alkyne for the preparation of 2,3,4,5-tetrasubstituted pyridine derivatives by three-component coupling reaction in the presence of Me3SiCl.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wim Schepens et al.
Organic letters, 8(19), 4247-4250 (2006-09-08)
A novel series of analogues of calcitriol (1) is developed featuring a spirocyclic central core resulting from C18/C21-connection and C15/C16-deletion (2a, 2b). The synthesis of the key intermediate involves an Eschenmoser rearrangement of an enantiomerically pure bromo-substituted cyclohexenol.
A highly efficient cycloaddition of vinylarenes with electron-deficient alkynes affording 1, 2-disubstituted-3, 4-dihydronaphthalenes catalysed by N, N-dimethylformamide dimethyl acetal
Jiang J-L, et al.
Organic & Biomolecular Chemistry, 5(12), 1854-1857 (2007)
Dimethylformamide dimethyl acetal as a building block in heterocyclic synthesis
Abu-Shanab FA, et al.
Journal of Heterocyclic Chemistry, 46(5), 801-827 (2009)
Synthesis of 1, 4-Diaryl-piperazine-2, 5-diones: New Behavior of N, N-Dimethylformamide Dimethyl Acetal (DMFDMA)
Abu-Shanab FA, et al.
Synthetic Communications, 38(3), 376-382 (2008)
Toshiaki Sasada et al.
The Journal of organic chemistry, 73(17), 6905-6908 (2008-07-30)
We have identified a Me3SiCl-mediated three-component coupling reaction of a functionalized enamine, N,N-dimethylformamide diethyl acetal, and an internal alkyne having an electron-withdrawing group that produces 2,3,4,5-tetrasubstituted pyridine derivatives in good to excellent yields via a single-step reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service