Skip to Content
Merck
All Photos(1)

Key Documents

239607

Sigma-Aldrich

Bromoethane

ReagentPlus®, ≥99%

Synonym(s):

Ethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Br
CAS Number:
Molecular Weight:
108.97
Beilstein:
1209224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
bp:
37-40 °C (lit.)
vapor pressure:
25.32 psi ( 55 °C)
7.54 psi ( 20 °C)

vapor density

~3.75 (vs air)

Quality Level

vapor pressure

25.32 psi ( 55 °C)
7.54 psi ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

952 °F

expl. lim.

11.25 %

refractive index

n20/D 1.425 (lit.)

bp

37-40 °C (lit.)

mp

−119 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCBr

InChI

1S/C2H5Br/c1-2-3/h2H2,1H3

InChI key

RDHPKYGYEGBMSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Inhibition of hydrogen-air mixtures by using chemical vapor additives: Compares the effectiveness of bromoethane and other brominated hydrocarbons in minimizing hydrogen hazards (SK Das et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Ozone 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave Spectrum, Structure, and Quadrupole Coupling Constant Tensor of Ethyl Bromide.
Flanagan C and Pierce L.
J. Chem. Phys. , 38(12), 2963-2969 (1963)
Microwave Spectra of Molecules Exhibiting Internal Rotation. V. Barrier Height in Ethyl Chloride and Ethyl Bromide.
Lide Jr DR.
J. Chem. Phys. , 30(1), 37-39 (1959)
J R Bucher et al.
Toxicology and applied pharmacology, 130(1), 169-173 (1995-01-01)
Chloroethane and bromoethane have been shown to cause a marked uterine tumor response in B6C3F1 mice exposed for 2 years. These chemicals are nearly unique in this regard among the nearly 400 chemicals studied by the National Toxicology Program, and
Paula Teper-Bamnolker et al.
Plant physiology, 158(4), 2053-2067 (2012-03-01)
Potato (Solanum tuberosum) tuber, a swollen underground stem, is used as a model system for the study of dormancy release and sprouting. Natural dormancy release, at room temperature, is initiated by tuber apical bud meristem (TAB-meristem) sprouting characterized by apical
Kenneth C Westaway et al.
The journal of physical chemistry. A, 112(41), 10264-10273 (2008-09-26)
Nucleophile (11)C/ (14)C [ k (11)/ k (14)] and secondary alpha-deuterium [( k H/ k D) alpha] kinetic isotope effects (KIEs) were measured for the S N2 reactions between tetrabutylammonium cyanide and ethyl iodide, bromide, chloride, and tosylate in anhydrous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service