Skip to Content
Merck
All Photos(3)

Key Documents

124052

Sigma-Aldrich

Bromoethane

reagent grade, 98%

Synonym(s):

Ethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Br
CAS Number:
Molecular Weight:
108.97
Beilstein:
1209224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
37-40 °C (lit.)
vapor pressure:
25.32 psi ( 55 °C)
7.54 psi ( 20 °C)

grade

reagent grade

Quality Level

vapor density

~3.75 (vs air)

vapor pressure

25.32 psi ( 55 °C)
7.54 psi ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

952 °F

expl. lim.

11.25 %

refractive index

n20/D 1.425 (lit.)

bp

37-40 °C (lit.)

mp

−119 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCBr

InChI

1S/C2H5Br/c1-2-3/h2H2,1H3

InChI key

RDHPKYGYEGBMSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromoethane (Ethyl bromide) is an alkyl halide. Microwave spectral investigations of ethyl bromide and its isotopic species have been reported. It has been synthesized either by employing Grignard waste water as a precursor3 or by reacting potassium bromide with concentrated sulfuric acid in ethanol.

Application

Bromoethane may be employed as a promoter during the preparation of 1-butene.4 It has been used to prepare anion-exchange membranes, which can be used for the isolation and purification of plasmid DNA and also for the separation of plasmid DNA and RNA from cell lysates, measured using ultraviolet-visible light spectrophotometer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Ozone 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramsden.N.E et al.
A-Level Chemistry (2000)
"Preparation of inorganic?organic anion-exchange membranes and their application in plasmid DNA and RNA separation"
Chang S-C, et al.
Journal of Membrane Science, 311(1), 336-348 (2008)
Microwave Spectrum, Structure, and Quadrupole Coupling Constant Tensor of Ethyl Bromide.
Flanagan C and Pierce L.
J. Chem. Phys. , 38(12), 2963-2969 (1963)
Paula Teper-Bamnolker et al.
Plant physiology, 158(4), 2053-2067 (2012-03-01)
Potato (Solanum tuberosum) tuber, a swollen underground stem, is used as a model system for the study of dormancy release and sprouting. Natural dormancy release, at room temperature, is initiated by tuber apical bud meristem (TAB-meristem) sprouting characterized by apical
Kenneth C Westaway et al.
The journal of physical chemistry. A, 112(41), 10264-10273 (2008-09-26)
Nucleophile (11)C/ (14)C [ k (11)/ k (14)] and secondary alpha-deuterium [( k H/ k D) alpha] kinetic isotope effects (KIEs) were measured for the S N2 reactions between tetrabutylammonium cyanide and ethyl iodide, bromide, chloride, and tosylate in anhydrous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service