Skip to Content
Merck
All Photos(1)

Key Documents

22192

Supelco

Cellulose Chromatography Resin

acetate phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C116H116O64
CAS Number:
Molecular Weight:
2534.12
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Product Name

Cellulose acetate phthalate,

form

powder

Quality Level

technique(s)

LPLC: suitable

matrix

acetylated cellulose

matrix active group

phthalate

capacity

1.0 meq/mL

separation technique

reversed phase

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1COC(=O)C)O)OC(=O)c4c(cccc4)C(=O)O)O)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)OC(=O)c3c(cccc3)C(=O)O)O)COC(=O)C

InChI

1S/C32H34O19/c1-13(33)45-11-19-21(35)25(49-29(42)17-9-5-3-7-15(17)27(38)39)23(37)32(48-19)51-24-20(12-46-14(2)34)47-31(44)26(22(24)36)50-30(43)18-10-6-4-8-16(18)28(40)41/h3-10,19-26,31-32,35-37,44H,11-12H2,1-2H3,(H,38,39)(H,40,41)/t19-,20-,21-,22+,23-,24-,25+,26-,31-,32+/m1/s1

InChI key

DMNFZTKVDDFCQE-WJXNTJCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose acetate phthalate (CAP) is a microbicide which inactivates sexually transmitted disease (STD) pathogens like HIV-1. CAP has been used for enteric film coating of tablets and capsules.

Application

CAP is used as a stabiling agent in the preparation of Pseudolatex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Robert Neurath et al.
BMC infectious diseases, 3, 27-27 (2003-11-18)
Cellulose acetate phthalate (CAP) has been used for several decades in the pharmaceutical industry for enteric film coating of oral tablets and capsules. Micronized CAP, available commercially as "Aquateric" and containing additional ingredients required for micronization, used for tablet coating
A Robert Neurath et al.
BMC infectious diseases, 2, 6-6 (2002-05-02)
Cellulose acetate phthalate (CAP), a promising candidate microbicide for prevention of sexual transmission of the human immunodeficiency virus type 1 (HIV-1) and other sexually transmitted disease (STD) pathogens, was shown to inactivate HIV-1 and to block the coreceptor binding site
Ju Hyeong Jeon et al.
Journal of periodontology, 79(8), 1457-1464 (2008-08-05)
The cholesterol-lowering drug simvastatin promotes bone formation in cell cultures and animal models. In previous studies, devices for the controlled, localized delivery of simvastatin hydroxyacid enhanced osteoblastic activity in vitro. The objective of this investigation was to determine bioactivity of
Chaobo Huang et al.
Biomaterials, 33(3), 962-969 (2011-10-25)
Despite many advances in modern medicine, human immunodeficiency virus (HIV) still affects the health of millions of people world-wide and much effort is put in developing methods to either prevent infection or to eradicate the virus after infection has occurred.
James C DiNunzio et al.
Molecular pharmaceutics, 5(6), 968-980 (2009-05-13)
Amorphous engineered particle compositions of itraconazole (ITZ) and potential concentration enhancing polymers, cellulose acetate phthalate (CAP) and polyvinyl acetate phthalate (PVAP), were produced by ultra-rapid freezing to investigate the effect of these polymers on the bioavailability of ITZ solid dispersions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service