Skip to Content
Merck
All Photos(1)

Key Documents

21170

Sigma-Aldrich

Calcium hydride

purum p.a., ≥97.0% (gas-volumetric), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2Ca
CAS Number:
Molecular Weight:
42.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
form:
powder

grade

purum p.a.

Quality Level

Assay

≥97.0% (gas-volumetric)

form

powder

reaction suitability

reagent type: reductant

mp

816 °C (lit.)

anion traces

chloride (Cl-): ≤2000 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Cd: ≤0.005%
Co: ≤0.005%
Cu: ≤0.02%
Fe: ≤0.005%
K: ≤0.01%
Na: ≤0.05%
Ni: ≤0.005%
Pb: ≤0.005%
Zn: ≤0.005%

SMILES string

[Ca]

InChI

1S/Ca.2H

InChI key

FAQLAUHZSGTTLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Calcium hydride can be used as a reducing agent for the dechlorination of chlorinated organic compounds. It can also be used as a drying agent to dry pyridine.

Other Notes

Desiccant for aprotic solvents, e.g. benzene, dioxane, DMF and HMPT

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desiccant efficiency in solvent drying. 3. Dipolar aprotic solvents.
D R Burfield et al.
The Journal of organic chemistry, 43(20), 3966-3968 (1978-09-01)
Juliane F Ferreira et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 189, 44-50 (2017-08-12)
A new quinoline-amide calix[4]arene 3-receptor for detection of hazardous anions and cations have been synthesized. The 3-receptor was examined for its sensing properties towards several different anions (Cr
Selective mechanochemical dehalogenation of chlorobenzenes over calcium hydride.
Loiselle S, et al.
Environmental Science & Technology, 31(1, 261-265 (1991)
Application of mechanical activation to decomposition of toxic chlorinated organic compounds
Korolev K, et al.
Chemistry for Sustainable Development, 11, 489-496 (2003)
D.R. Burfield et al.
The Journal of Organic Chemistry, 42, 3060-3060 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service