Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium hydride
Synonym(s):
Calcium hydride
Empirical Formula (Hill Notation):
H
2
Ca
CAS Number:
7789-78-8
Molecular Weight:
42.09
EC Number:
232-189-2
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (5)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
208027
Calcium hydride,
reagent grade, 95% (gas-volumetric)
Expand
View Pricing
8.02100
Calcium hydride,
for synthesis
Expand
View Pricing
558257
Calcium hydride,
99.9% trace metals basis
Expand
View Pricing
213268
Calcium hydride,
powder, 0-2 mm, reagent grade, ≥90% (gas-volumetric)
Expand
View Pricing
497355
Calcium hydride,
powder, 99.99% trace metals basis
Expand
View Pricing
21170
Calcium hydride,
purum p.a., ≥97.0% (gas-volumetric), powder
Expand
View Pricing