Skip to Content
Merck
All Photos(1)

Documents

21059

Supelco

Calcium ion solution for ISE

0.1 M Ca, analytical standard (for ion-selective electrodes)

Synonym(s):

Calcium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

analytical standard (for ion-selective electrodes)
for ion-selective electrodes

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

0.1 M Ca

SMILES string

Cl[Ca]Cl

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium ion solution (CaCl2) contains dissolved calcium ions and chloride ions.
Visit our Sensor Applications portal to learn more.

Application

CaCl2 solution was used in the photolysis of caged compounds while obtaining patch-clamp, in an experiment conducted to study olfactory transduction in dissociated mouse olfactory sensory neurons.

Preparation Note

prepared with CaCl2.2H2O and H2O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rabia Iqbal et al.
Probiotics and antimicrobial proteins, 11(3), 830-839 (2018-03-14)
Longevity of probiotic is the main concern for getting maximum benefits when added in food product. Bifidobacterium, a probiotic, tends to lose its viability during gastrointestinal track (GIT) transit and storage of food. Their viability can be enhanced through microencapsulation
Anna Boccaccio et al.
Journal of visualized experiments : JoVE, (55)(55), e3195-e3195 (2011-11-09)
Photolysis of caged compounds allows the production of rapid and localized increases in the concentration of various physiologically active compounds. Caged compounds are molecules made physiologically inactive by a chemical cage that can be broken by a flash of ultraviolet
John S. Holman, Phil Stone
Chemistry (Weinheim An Der Bergstrasse, Germany), 170-170 (2001)
Rachel Deplus et al.
Nucleic acids research, 42(13), 8285-8296 (2014-06-25)
DNA methylation is a central epigenetic modification in mammals, with essential roles in development and disease. De novo DNA methyltransferases establish DNA methylation patterns in specific regions within the genome by mechanisms that remain poorly understood. Here we show that
Konstantinos Malliaras et al.
EMBO molecular medicine, 6(6), 760-777 (2014-05-07)
Controversy surrounds the identity, origin, and physiologic role of endogenous cardiomyocyte progenitors in adult mammals. Using an inducible genetic labeling approach to identify small non-myocyte cells expressing cardiac markers, we find that activated endogenous cardioblasts are rarely evident in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service