Skip to Content
Merck

17248

N-Decanoyl-DL-homoserine lactone

≥97.0% (HPLC)

Synonym(s):

N-(tetrahydro-2-oxo-3-furanyl)-Decanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H25NO3
CAS Number:
Molecular Weight:
255.35
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Beilstein/REAXYS Number:
1644700
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-Decanoyl-DL-homoserine lactone, ≥97.0% (HPLC)

Quality Segment

assay

≥97.0% (HPLC)

form

powder

color

white

mp

111-114 °C

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

CCCCCCCCCC(=O)NC1CCOC1=O

InChI

1S/C14H25NO3/c1-2-3-4-5-6-7-8-9-13(16)15-12-10-11-18-14(12)17/h12H,2-11H2,1H3,(H,15,16)

InChI key

TZWZKDULKILUPV-UHFFFAOYSA-N

Application

Application test: Induces violacein expression in a Chromobacterium violaceum mutant usually not able to produce homoserine lactones

Biochem/physiol Actions

N-Decanoyl-DL-homoserine lactone (N-C10-HSL) is among a group of homoserine lactones that includes; N-octanoyl-homoserine lactone (N-C8-HSL), N-(3-oxodecanoyl) homoserine-L-lactone (3-oxo-C10 HSL), N-(3-oxododecanoyl)homoserine-L-lactone (3-oxo-C12-HSL), N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-oxo-C14-HSL, N-(3-hydroxydecanoyl)-L-homoserine lactone, and N-(3-hydroxyoctanoyl)-L-homoserine lactone involved in the processes of bacterial quorum sensing. These N-acyl-homoserine lactones are used to study the processes and mechanisms of bacterial quorum sensing.
N-Decanoyl-DL-homoserine lactone is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such as Echerichia and Salmonella are involved in quorum sensing, cell to cell communication among bacteria. Some AHLs are potent chemoattractants for human immune cells such as neutrophils.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
17248-25MG04061838751638