200905
Magnesium
turnings, reagent grade, 98%
Synonym(s):
Magnesium element
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
grade
reagent grade
Quality Level
vapor pressure
1 mmHg ( 621 °C)
Assay
98%
form
turnings
autoignition temp.
950 °F
reaction suitability
reagent type: reductant
resistivity
4.46 μΩ-cm, 20°C
bp
1090 °C (lit.)
mp
648 °C (lit.)
density
1.74 g/mL at 25 °C (lit.)
SMILES string
[Mg]
InChI
1S/Mg
InChI key
FYYHWMGAXLPEAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Magnesium turnings are generally used in the preparation of various organomagnesium compounds such as Grignard reagents. It can also be used to catalyze Pinacol coupling and Reformatsky reaction.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Self-heat. 1 - Water-react 2
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Magnesium-induced pinacol coupling of aromatic aldehydes and ketones in water under ultrasound.
Indian Journal of Chemistry, 43B(6), 196?198-196?198 (2004)
The Grignard Reagents.
Organometallics, 28(6), 1598-1605 (2009)
A Simple and Efficient Procedure of Low Valent Iron-or Copper-Mediated Reformatsky Reaction of Aldehydes.
The Journal of Organic Chemistry, 72(24), 9357-9359 (2007)
Grignard reagent formation.
Coordination Chemistry Reviews, 248(7-8), 623-652 (2004)
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service