Skip to Content
Merck
All Photos(1)

Documents

723320

Sigma-Aldrich

Magnesium

in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 621 °C)

form

turnings

autoignition temp.

950 °F

composition

anhydrous tetrahydrofuran, 37.5 mmol

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium turnings (Mg) can be used to prepare reactive organomagnesium compounds (Grignard reagents, GR) by reacting with alkyl, allyl, vinyl, or aryl halides. These GR reagents are widely used in organic synthesis to produce useful chemical intermediates. Mg can also be used to prepare tetraisopropyl- and tetrabutyl-distibane by dehalogenation of diisopropyl- and dibutyl-antinomy bromide, respectively.

Other Notes

Based on preparing 0.5 M solution of organomagnesium reagent and 40% headspace.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Self-heat. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reduktion von isopropyl-und butylantimonbromiden mit magnesium
Breunig HJ and KW
Journal of Organometallic Chemistry, 186(1), C5-C8 (1980)
Titanium-magnesium-assisted scission of 1, 4-bis (trimethylsilyl)-1, 3-butadiyne: synthesis and structure of a titanium (III) tweezer complex,[(. eta. 5-C5HMe4) 2Ti (. eta. 1-C. tplbond. CSiMe3) 2][Mg (THF) Cl]
Troyanov SI, et al.
Organometallics, 12(7), 2820-2824 (1993)
Mechanical activation of magnesium turnings for the preparation of reactive Grignard reagents
Baker KV, et al.
The Journal of Organic Chemistry, 56(2), 698-703 (1991)
Shuvendu Biswas et al.
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate
Benjamin Chaigne-Delalande et al.
Science (New York, N.Y.), 341(6142), 186-191 (2013-07-13)
The magnesium transporter 1 (MAGT1) is a critical regulator of basal intracellular free magnesium (Mg(2+)) concentrations. Individuals with genetic deficiencies in MAGT1 have high levels of Epstein-Barr virus (EBV) and a predisposition to lymphoma. We show that decreased intracellular free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service