Skip to Content
Merck
All Photos(1)

Documents

12279

Supelco

1,2-Propanediol

analytical standard

Synonym(s):

1,2-PDO, Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E490
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

779 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

12.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.432 (lit.)
n20/D 1.433

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Propanediol is a compound found to be chemically similar to ethylene glycol and is widely used in automobile antifreeze, activated charcoal preparations, cosmetics, beverages, ointments, etc. It is used as a diluent for the intravenous preparations of poorly water-soluble drugs.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

1,2-Propanediol may be used as an analyte protectant for the gas chromatography-mass spectrometric detection of environmental contaminants in honey, honeybees and pollens. It is also used as an analytical reference standard for the quantification of the analyte in biological fluids using headspace-gas chromatography (HS-GC) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid analysis of plasma/serum ethylene and propylene glycol by headspace gas chromatography
Ehlers A, et al.
SpringerPlus, 2(1), 203-203 (2013)
Multi-residue analysis of 80 environmental contaminants in honeys, honeybees and pollens by one extraction procedure followed by liquid and gas chromatography coupled with mass spectrometric detection.
Wiest L, et al.
Journal of Chromatography A, 1218(34), 5743-5756 (2011)
Gai Ling Li et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(1-2), 49-57 (2002-10-26)
To achieve a therapeutical effect of the anti-Parkinson's drug R-apomorphine via iontophoresis delivery, enhancement strategies in vitro were explored using three structurally related enhancers, lauric acid (LA), dodecyltrimethylammonium bromide (DTAB) and Laureth-3 oxyethylene ether (C(12)EO(3)). Human stratum corneum and shed
G A Reichard et al.
Diabetes, 35(6), 668-674 (1986-06-01)
Plasma acetone turnover rates were measured with the primed continuous infusion of 2-[14C]acetone in patients with moderate to severe diabetic ketoacidosis. Plasma acetone turnover rates ranged from 1.52 to 15.9 mumol X kg-1 X min-1 (108-1038 mumol X 1.73 m-2
R S Rajasenan et al.
Toxicology and applied pharmacology, 135(1), 89-99 (1995-11-01)
Engrafting components of human immune systems in severe combined immunodeficient (SCID) mice has been utilized to investigate the pathogenesis of several human autoimmune diseases and may provide a model for studying idiosyncratic drug toxicity. The purpose of this investigation was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service