Skip to Content
Merck
All Photos(3)

Documents

07672

Sigma-Aldrich

N,N-Diethyl-p-phenylenediamine sulfate salt

≥98.0% (T)

Synonym(s):

4-Amino-N,N-diethylaniline sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H4NH2 · H2SO4
CAS Number:
Molecular Weight:
262.33
Beilstein:
4219768
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

solid

mp

184-186 °C (lit.)
184-187 °C

SMILES string

OS(O)(=O)=O.CCN(CC)c1ccc(N)cc1

InChI

1S/C10H16N2.H2O4S/c1-3-12(4-2)10-7-5-9(11)6-8-10;1-5(2,3)4/h5-8H,3-4,11H2,1-2H3;(H2,1,2,3,4)

InChI key

AYLDJQABCMPYEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Diethyl-p-phenylenediamine sulfate salt has been used to prepare N,N-diethyl-p-phenylenediamine (DPD) solution, which is commonly used as a chromogenic indicator for the determination of free and total chlorine by titrimetric, colorimetric and spectroscopic methods.{50

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved N, N?-diethyl-p-phenylenediamine (DPD) method for the determination of free chlorine based on multiple wavelength detection.
Moberg L & Karlberg B
Analytica Chimica Acta, 407(1-2), 127-133 (2000)
Improvements in the N, N-diethyl-p-phenylenediamine method for the determination of free and combined residual chlorine through the use of FIA.
Gordon G, et al.
Talanta, 38(2), 145-149 (1991)
Masoumeh Hasani et al.
Journal of hazardous materials, 157(1), 161-169 (2008-02-15)
A multicomponent analysis method based on principal component analysis-artificial neural network models (PC-ANN) is proposed for the determination of phenolic compounds. The method relies on the oxidative coupling of phenols (phenol, 2 chlorophenol, 3-chlorophenol and 4-chlorophenol) to N,N-diethyl-p-phenylenediamine in the
Jessica A Kershaw et al.
Analytical and bioanalytical chemistry, 379(4), 707-713 (2004-06-04)
The effect of Cu(II) on the determination of homocysteine via its electrochemically initiated reaction with N, N-diethyl-p-phenylenediamine is examined. The presence of copper inhibited the detection process for homocysteine owing to a complexation reaction occurring. This provided an indirect route
Z B Ogel et al.
Applied microbiology and biotechnology, 71(6), 853-862 (2006-01-04)
Scytalidium thermophilum produces an extracellular phenol oxidase on glucose-containing medium. Certain phenolic acids, specifically gallic acid and tannic acid, induce the expression of the enzyme. Production at 45 degrees C in batch cultures is growth-associated and is enhanced in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service