Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N,N
-Diethyl-
p
-phenylenediamine sulfate salt
Synonym(s):
4-Amino-
N,N
-diethylaniline sulfate salt
Linear Formula:
(C
2
H
5
)
2
NC
6
H
4
NH
2
· H
2
SO
4
CAS Number:
6283-63-2
Molecular Weight:
262.33
EC Number:
228-500-6
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
07672
N,N
-Diethyl-
p
-phenylenediamine sulfate salt,
≥98.0% (T)
Expand
View Pricing
07670
N,N
-Diethyl-
p
-phenylenediamine sulfate salt,
for spectrophotometric det. of S
2-
, Cl
2
, ≥99.0%
Expand
View Pricing