Skip to Content
Merck
All Photos(2)

Key Documents

06813

Supelco

α-Boswellic acid

analytical standard

Synonym(s):

(3α,4β)-3-Hydroxyolean-12-en-23-oic acid, 3α-Hydroxyolean-12-en-24-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
Beilstein:
3074442
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)CC[C@]2(C)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@@H](O)[C@@](C)([C@@H]5CC[C@@]34C)C(O)=O)[C@@H]2C1

InChI

1S/C30H48O3/c1-25(2)14-15-26(3)16-17-28(5)19(20(26)18-25)8-9-21-27(4)12-11-23(31)30(7,24(32)33)22(27)10-13-29(21,28)6/h8,20-23,31H,9-18H2,1-7H3,(H,32,33)/t20-,21+,22+,23+,26+,27+,28+,29+,30+/m0/s1

InChI key

BZXULBWGROURAF-IKNLXHIFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Boswellic acid is a pentacyclic triterpene acid belonging to the genus Boswellia. It is a biologically active constituent of Boswellia serrata, known for its anti-inflammatory activity.

Application

α-Boswellic acid may be used as an analytical reference standard for the quantification of the analyte in the following:        
  • Oleogum resins of different Boswellia species using high-performance liquid chromatography with tandem mass spectrometric detection (HPLC-MS/MS).        
  • Frankincense extracts of different Boswellia species using two-dimensional liquid chromatography electrospray ionization tandem mass spectrometry (LC/LC/ESI-MS/MS).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative analysis of pentacyclic triterpenic acid compositions in oleogum resins of different Boswellia species and their in vitro cytotoxicity against treatment-resistant human breast cancer cells
Schmiech M, et al.
Molecules (Basel), 24(11), 2153-2153 (2019)
Analysis of frankincense from various Boswellia species with inhibitory activity on human drug metabolising cytochrome P450 enzymes using liquid chromatography mass spectrometry after automated on-line extraction
Frank A and Unger M
Journal of Chromatography A, 1112(1-2), 255-262 (2006)
Comparison Between HPLC and HPTLC Densitometry for the Determination of 11-keto-?-boswellic acid and 3-acetyl-11-keto-?-boswellic acid from Boswellia serrata Extract
Mehta M, et al.
Indian Journal of Pharmaceutical Education and Research, 50(3), 418-423 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service