Skip to Content
Merck

A9855

3-Acetyl-11-keto-β-boswellic acid

from Boswellia serrata

Synonym(s):

AKBA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H48O5
CAS Number:
Molecular Weight:
512.72
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Boswellia serrata

Quality Level

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O([C@H]1[C@]([C@H]2[C@@]([C@@H]3[C@]([C@@]4(CC[C@@]5([C@@H]([C@H]([C@@H](CC5)C)C)C4=CC3=O)C)C)(CC2)C)(CC1)C)(C)C(=O)O)C(=O)C

InChI

1S/C32H48O5/c1-18-9-12-28(4)15-16-30(6)21(25(28)19(18)2)17-22(34)26-29(5)13-11-24(37-20(3)33)32(8,27(35)36)23(29)10-14-31(26,30)7/h17-19,23-26H,9-16H2,1-8H3,(H,35,36)/t18-,19+,23-,24-,25+,26-,28-,29+,30-,31-,32-/m1/s1

InChI key

HMMGKOVEOFBCAU-BCDBGHSCSA-N

General description

Acetyl-11-keto-β-boswellic acid (AKBA) is extracted from the resin of Boswellia serrata.

Application

3-Acetyl-11-keto-β-boswellic acid (AKBA) has been used:
  • to test its effects as an anti-proliferating agent on hepatic stellate cells (HSCs) proliferation and synergism in combination with imatinib in in vitro models
  • to study its anti-osteoporotic activity on ovariectomy-induced osteoporosis in female Sprague Dawley rats
  • in a comparative study with aspirin to test its effects as an anti-inflammatory drug to prevent intestinal adenomatous polyposis in APCMin/+ mice

Biochem/physiol Actions

Acetyl-11-keto-β-boswellic acid (AKBA) displays antibacterial and anti-biofilm activities against Staphylococcus epidermidis and S. aureus, respectively. It is an activator of nuclear factor erythroid-2-related factor 2 (Nrf2) and exerts neuroprotective properties against ischemic injury in both in vitro and in vivo studies.
Acetyl-11-keto-beta-boswellic acid is a pentacyclic triterpene that inhibits 5-lipoxygenase in a selective, enzyme-directed, nonredox, and noncompetitive manner. It has also been shown to inhibit topoisomerase I and NF-κB signaling.


Still not finding the right product?

Explore all of our products under 3-Acetyl-11-keto-β-boswellic acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Alsaba F Raja et al.
BMC microbiology, 11, 54-54 (2011-03-17)
Boswellic acids are pentacyclic triterpenes, which are produced in plants belonging to the genus Boswellia. Boswellic acids appear in the resin exudates of the plant and it makes up 25-35% of the resin. β-boswellic acid, 11-keto-β-boswellic acid and acetyl-11-keto-β-boswellic acid
Mona Abdel-Tawab et al.
Clinical pharmacokinetics, 50(6), 349-369 (2011-05-11)
Non-steroidal anti-inflammatory drug (NSAID) intake is associated with high prevalence of gastrointestinal or cardiovascular adverse effects. All efforts to develop NSAIDs that spare the gastrointestinal tract and the cardiovasculature are still far from achieving a breakthrough. In the last two
Ruiqi Wang et al.
Drug discoveries & therapeutics, 8(1), 25-32 (2014-03-22)
Acetyl-11-keto-beta-BA (AKBA), a component of the gum resin of Boswellia serrata, has been recognized as a promising agent for the prevention of intestinal tumorigenesis. Aspirin, a non-steroidal anti-inflammatory drug (NSAID), has also been considered to have the activity against intestinal



Global Trade Item Number

SKUGTIN
A9855-5MG04061832860831