Skip to Content
Merck
All Photos(1)

Key Documents

03550

Sigma-Aldrich

Ethylenediamine

puriss. p.a., absolute, ≥99.5% (GC)

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605263
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.07 (vs air)

Quality Level

vapor pressure

10 mmHg ( 20 °C)

grade

absolute
puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

716 °F

expl. lim.

16 %

impurities

≤0.5% water

refractive index

n20/D 1.456
n20/D 1.4565 (lit.)

bp

118 °C (lit.)

mp

8.5 °C (lit.)

solubility

H2O: soluble at 

density

0.899 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

functional group

amine

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Gene Information

human ... FNTA(2339)

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediamine is a linear aliphatic diamine, widely used as a building block in organic synthesis. It readily forms heterocyclic imidazolidine derivatives, because of its bifunctional nature, having two amines. Ethylenediamine is also utilized as a raw material for the synthesis of chelating agents, polymers, agrochemicals and pharmaceutical intermediates.

Application

Ethylenediamine can be employed as:
  • A reactant in the synthesis of water-swellable polyurethane urea gels by copolymerization.
  • A solvent along with dodecanethiol in the preparation of CdS nanowires by solvothermal method.
  • A chelating agent in the synthesis of β-Co(OH)2 nanocrystals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvothermal synthesis of CdS nanowires in a mixed solvent of ethylenediamine and dodecanethiol
Xu Dan, et al.
The Journal of Physical Chemistry B, 109(30), 14344-14349 (2005)
Low temperature route towards new materials: solvothermal synthesis of metal chalcogenides in ethylenediamine
Li J, et al.
Coordination Chemistry Reviews, 190, 707-735 (1999)
Amines, aliphatic
Eller K, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Synthesis, cytotoxicity, and antileishmanial activity of N, N'-disubstituted ethylenediamine and imidazolidine derivatives
de Carvalho GSG, et al.
TheScientificWorldJournal, 10(51), 1723-1730 (2010)
Concise solid-phase synthesis of inverse poly (amidoamine) dendrons using AB 2 building blocks
Huang A Y-T, et al.
Chemical Communications (Cambridge, England), 49(51), 5784-5786 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service