Skip to Content
Merck
All Photos(2)

Key Documents

E1521

Sigma-Aldrich

Ethylenediamine

BioXtra

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605263
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030201
PubChem Substance ID:
NACRES:
NA.21
bp:
118 °C (lit.)
vapor pressure:
10 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.07 (vs air)

vapor pressure

10 mmHg ( 20 °C)

product line

BioXtra

Assay

>99%

form

liquid

autoignition temp.

716 °F

expl. lim.

16 %

impurities

≤0.0005% Phosphorus (P)

ign. residue

≤0.1%

refractive index

n20/D 1.4565 (lit.)

pH

12.2 (20 °C, 110 g/L)

bp

118 °C (lit.)

mp

8.5 °C (lit.)

solubility

H2O: soluble at 

density

0.899 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

functional group

amine

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Gene Information

human ... FNTA(2339)

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediamine (en) is a linear aliphatic diamine that can be synthesized by reacting ethanolamine and ammonia in the presence of zeolite catalyst.[1] Its effect as an allergen has been investigated.[2] It participates in the synthesis of metal chalcogenides[3] and thiogallates.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minjeong Park et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-10)
Cu nanowires (NWs) possess remarkable potential a slow-cost heat transfer material in modern electronic devices. However, Cu NWs with high aspect ratios undergo surface oxidation, resulting in performance degradation. A growth temperature of approximately <1000 °C is required for preventing
Paz Vaqueiro
Inorganic chemistry, 45(10), 4150-4156 (2006-05-09)
Five new thiogallates have been prepared solvothermally in the presence of ethylenediamine and characterized by single-crystal X-ray diffraction, thermogravimetry, and elemental analysis. [enH2][Ga4S7(en)2] (1), which crystallizes in the monoclinic space group P2(1)/c with lattice parameters a = 12.8698(12) angstroms, b
Effect of dealumination of mordenite by acid leaching for selective synthesis of ethylenediamine from ethanolamine.
Segawa K and Shimura T.
Applied Catalysis A: General, 194, 309-317 (2000)
S D Prystowsky et al.
Archives of dermatology, 115(8), 959-962 (1979-08-01)
A study population of 1,158 paid adult volunteers was obtained. Prior to patch testing, a history of previous exposure to four allergens also was obtained. Prevalence of positive reactions to patch tests was nickel, 5.8%; neomycin, 1.1%; ethylenediamine, 0.43%; and
Yadong Li et al.
Inorganic chemistry, 38(21), 4737-4740 (2001-10-24)
Reaction of sulfur and selenium with elemental or ionic metals in liquid ethylenediamine at room temperature produced metal chalcogenides ME (M = Cu, Sn, Co(2+), Ni(2+), Mn(2+), or Pb(2+); E = S or Se), Ag(2)E, M(2)E(3) (M = Bi or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service