Skip to Content
Merck
All Photos(1)

Documents

01338

Supelco

Bergamottin

analytical standard

Synonym(s):

Bergamotine, Bergaptin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
Beilstein:
1353914
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

75-80 °C

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC\C(C)=C\COc1c2C=CC(=O)Oc2cc3occc13

InChI

1S/C21H22O4/c1-14(2)5-4-6-15(3)9-11-24-21-16-7-8-20(22)25-19(16)13-18-17(21)10-12-23-18/h5,7-10,12-13H,4,6,11H2,1-3H3/b15-9+

InChI key

DBMJZOMNXBSRED-OQLLNIDSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bergamottin, a member of the furanocoumarin family, is primarily found in grapefruit juice. It is reported to be a novel signal transducer and activator of the transcription 3 (STAT3) inhibitor. It is a known drug inhibitor of cytochrome P450 34A and also exhibits diverse anti-tumor and anti-inflammatory activities.

Application

Bergamottin may be used as an analytical standard to test its efficacy in the following:
  • Blocker of STAT3 activation pathway in tumor cells; its potential in the therapy of multiple myeloma (MM) and other cancers is investigated.
  • Prevention and the treatment of cancer via suppression of the NF-kB signaling pathway and its regulated gene products.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anastasia Messer et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(12), 3224-3231 (2011-09-29)
Furocoumarins are phototoxic and photogenotoxic natural plant constituents occurring in cosmetics, food and drugs. Grapefruit juice is considered as a major dietary source of furocoumarins although few is known about the variability of furocoumarins in grapefruit juice. We analyzed the
Paul F Cancalon et al.
Journal of food science, 76(4), C543-C548 (2012-03-16)
Since the early 1990's, grapefruit juice has been implicated in drug interaction with various furanocoumarins (FCs) now associated with the effect. Although FCs are present in various fruits and vegetables, it is their presence in grapefruit that has attracted the
Yong Pil Hwang et al.
Molecular nutrition & food research, 54(7), 977-990 (2009-11-28)
Matrix metalloproteinase (MMP) plays an important role in the invasion and metastasis of cancer cells. The inhibitory effects of bergamottin, a cytochrome P450 inhibitor from Citrus paradis (grapefruit), on tumor invasion and migration and the possible mechanisms involved in this
Simvastatin in combination with bergamottin potentiates TNF-induced apoptosis through modulation of NF-?B signalling pathway in human chronic myelogenous leukaemia
Kim M-S, et al.
Pharmaceutical biology, 54(10), 2050-2060 (2016)
Y Uesawa et al.
Die Pharmazie, 66(7), 525-528 (2011-08-05)
Grapefruit juice (GFJ) is known to affect the pharmacokinetics of a variety of drugs administered concomitantly and this is due to inhibition of intestinal CYP3A, a barrier protein for drug absorption. Some compounds such as furanocoumarin derivatives have been reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service