
Bergamottin

Synonym(s):
Bergamotine, Bergaptin
Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing