Skip to Content
Merck
All Photos(2)

Key Documents

00290

Supelco

(−)-Sinigrin hydrate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16KNO9S2 · xH2O
CAS Number:
Molecular Weight:
397.46 (anhydrous basis)
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥99.0% (TLC)

optical activity

[α]/D -17.0±1°, c = 1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤6.0% water

mp

128 (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

[K+].[H]O[H].OC[C@H]1O[C@@H](S\C(CC=C)=N\OS([O-])(=O)=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C10H17NO9S2.K.H2O/c1-2-3-6(11-20-22(16,17)18)21-10-9(15)8(14)7(13)5(4-12)19-10;;/h2,5,7-10,12-15H,1,3-4H2,(H,16,17,18);;1H2/q;+1;/p-1/b11-6+;;/t5-,7-,8+,9-,10+;;/m1../s1

InChI key

IUBVMJHASFBYGW-WBMBWNLZSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A β-D-thioglucopyranoside occurring in black mustard seeds and horseradish root. Substrate for thioglucosidase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Morgan et al.
Chemico-biological interactions, 310, 108739-108739 (2019-07-10)
Phenol red (PR) is the standard pH indicator in various cell and tissue culture media, as it provides a quick check for the health of the culture. PR has also been used in multiple protocols to detect cellular hydrogen peroxide
Marta Lara-Lledó et al.
International journal of food microbiology, 156(1), 25-31 (2012-03-20)
The ability of Listeria (L.) monocytogenes to convert glucosinolates into antimicrobial isothiocyanates was investigated. Mustard glucosinolates in pure (sinigrin) or extract forms (sinigrin, oriental; sinalbin, yellow mustard) were used in broth media and in a polyvinyl polyethylene glycol graft copolymer
Monika Okulicz
Plant foods for human nutrition (Dordrecht, Netherlands), 65(3), 217-224 (2010-09-03)
Sinigrin (SIN) and allyl isothiocyanate (AITC) are compounds found in high concentrations in Brassica family vegetables, especially in Brussels sprouts. Recently, they have been used as a nutrition supplement for their preventive and medicinal effect on some types of cancer
Jóska Gerendás et al.
Journal of agricultural and food chemistry, 57(9), 3837-3844 (2009-03-25)
Food derived from Brassica species is rich in glucosinolates. Hydrolysis of these compounds by myrosinase yields isothiocyanates and other breakdown products, which due to their pungency represent the primary purpose of Indian mustard cultivation. Strong interactive effects of S (0.0
S Herzallah et al.
Journal of food protection, 74(12), 2162-2168 (2011-12-22)
The glucosinolate sinigrin (SNG) is converted by endogenous plant myrosinase or by bacterial myrosinase-like activity to form the potent antimicrobial allyl isothiocyanate. In order to use SNG as a natural antimicrobial precursor in food, it became important to better understand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service