Skip to Content
Merck
All Photos(2)

Key Documents

11585762001

Roche

DIG Wash and Block Buffer Set

greener alternative

storage temp.:2-8°C

Synonym(s):

wash and block buffer set, dig

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105326

Quality Level

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

Except for the Blocking solution, the 10x concentrated buffers are diluted to the desired concentrations with sterile double-distilled water (not included).
Washing, blocking and detection buffers (10x concentrated) for the immunological detection of DIG - (digoxigenin-)labeled probes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to using radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Application

The DIG Wash and Block Buffer Set is used at various steps of DIG hybridization and DIG detection.

Packaging

1 set containing 4 components.

Preparation Note

Working solution: dATP, dGTP, dTTP mixture:
For one labeling reaction pipette:
1 μl dATP, (vial 2)
1 μl dGTP, (vial 4)
1 μl dTTP, (vial 5)
to a reaction vial.

Note: If the same type of labeled deoxyribonucleoside-triphosphate is used repeatedly, we recommend the preparation of a mixture of equal parts of the other three deoxyribonucleoside-triphosphates for convenience.
Storage conditions (working solution):
  • After the first usage, the buffers should be stored further at 2 to 8 °C. We recommend to store the concentrated Blocking solution in aliquots at -15 to -25 °C.
  • When Autoclaved: Stock solution can be stored for several days to a week either unopened at RT or at 2 to 8 °C after opening. Alternatively, it can be stored in aliquots at -15 to -25 °C for up to 6 months.

Analysis Note

The buffers are autoclaved, filtered and tested for the absence of DNases and RNases according to the current Quality Control procedures. The buffers are function tested according to the standard procedure of the DIG Nucleic Acid Detection Kit.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Washing Buffer 10x concentrated

  • Maleic acid Buffer 10x concentrated

  • Blocking Solution 10x concentrated

  • Detection Buffer 10x concentrated

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicabledoes not flash

Flash Point(C)

Not applicabledoes not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiho Torii et al.
Cell reports, 35(3), 109014-109014 (2021-04-12)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has been identified as the causative agent of coronavirus disease 2019 (COVID-19). Although multiple mutations have been observed in SARS-CoV-2, functional analysis of each mutation of SARS-CoV-2 has been limited by the lack
Dan Sun et al.
Oncology letters, 20(4), 8-8 (2020-08-11)
Since human bladder cancer (BC) is a common malignancy of the urinary system with poor prognosis, it is crucial to clarify the molecular mechanisms of BC development and progression. To the best of our knowledge, the current study demonstrated for
James A Baum et al.
Journal of economic entomology, 105(2), 616-624 (2012-05-23)
The plant bugs Lygus hesperus Knight (Hemiptera: Miridae) and L. lineolaris (Palisot de Beauvois) have emerged as economic pests of cotton in the United States. These hemipteran species are refractory to the insect control traits found in genetically modified commercial
Siqian Feng et al.
Genetics, 217(3) (2021-03-28)
We describe a simple and efficient technique that allows scarless engineering of Drosophila genomic sequences near any landing site containing an inverted attP cassette, such as a MiMIC insertion. This two-step method combines phiC31 integrase-mediated site-specific integration and homing nuclease-mediated
James V McCann et al.
Genesis (New York, N.Y. : 2000), 58(7), e23369-e23369 (2020-06-17)
Extracellular vesicles (EVs) are abundant, lipid-enclosed vectors that contain nucleic acids and proteins, they can be secreted from donor cells and freely circulate, and they can be engulfed by recipient cells thus enabling systemic communication between heterotypic cell types. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service