Skip to Content
Merck

CSPD-RO

Roche

CSPD ready-to-use

Disodium 3-(4-methoxyspiro {1,2-dioxetane-3,2′-(5′-chloro)tricyclo [3.3.1.13,7]decan}-4-yl)phenyl phosphate

Synonym(s):

ready-to-use, cspd

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
41105500
NACRES:
NA.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

>98% (HPLC)

form

solution

mol wt

Mr 461

packaging

pkg of 1 mL (11655884001[25mM]), pkg of 2 × 50 mL (11755633001[0.25mM] ready-to-use)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

CSPD is a chemiluminescent substrate for alkaline phosphatase that enables extremely fast and sensitive detection of biomolecules by producing visible light. Light emission is recorded on X-ray film or on luminescence imager systems.

Application

CSPD ready-to-use has been used for:
  • the nonradioactive detection of labeled nucleic acid probes on membrane blots (Southern blot, Northern blot, Western blot and Dot blot)
  • colony and plaque hybridization
  • gel shift assay
  • sequencing
It may be used for the detection of alkaline phosphatase and alkaline phosphatase conjugates either in solution or on solid supports.

Features and Benefits

  • Save time with a fast, highly sensitive substrate.
  • Easily strip and reprobe membranes.
  • Take multiple exposures of up to two days for each experiment.
  • Reuse the substrate by filtering and storing in sodium azide.

Physical form

25mM solution (11.6mg/ml), 100x concentrated, colorless solution (11655884001).

0.25mM solution (0.116mg/ml), 1x concentrated, colorless solution (11755633001).

Preparation Note

Storage conditions (working solution): Washing Buffer: 15 to 25 °C
Maleic Acid Buffer: 15 to 25 °C
Detection Buffer: 15 to 25 °C
Blocking Stock Solution, 10x: 2 to 8 °C
Blocking Solution: always prepare fresh
Antibody Solution: 2 to 8 °C

Other Notes

For life science research only. Not for use in diagnostic procedures.
Purity: Purified by HPLC
Development time: Variable
Note: First exposure to X-ray film should be 15 to 30 minutes. Examine film and then determine other exposure times based on initial result.
Activated by preincubation (10 minutes at +37°C).
Apply substrate quickly to avoid drying of membrane (background).
Do not use plastic wrap to cover blot; use hybridization bags, acetate sheet protectors, or two sheets of transparent film.
Do not use nitrocellulose membranes.
Repeat exposures can be made several days after addition of substrate.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Martin et al.
BioTechniques, 18(5), 908-913 (1995-05-01)
Quantitative PCR and reverse transcription PCR (RT-PCR) are widely used in biomedical, industrial and other research applications to determine the number of RNA or DNA molecules of a specific type and/or sequence in a sample of interest. We have developed
Hoa Mai Nguyen et al.
Plant physiology, 163(2), 914-928 (2013-08-21)
The ω-3 polyunsaturated fatty acids account for more than 50% of total fatty acids in the green microalga Chlamydomonas reinhardtii, where they are present in both plastidic and extraplastidic membranes. In an effort to elucidate the lipid desaturation pathways in
Diane M Ramos et al.
BMC developmental biology, 6, 55-55 (2006-11-23)
Precise temporal and spatial regulation of transgene expression is a critical tool to investigate gene function in developing organisms. The most commonly used technique to achieve tight control of transgene expression, however, requires the use of specific DNA enhancers that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service