Skip to Content
Merck

02-102

Endothelial Cell Growth Supplement, 150mg

The Endothelial Cell Growth Supplement is designed for use as a cell culture media supplement. This cell growth supplement is available in a 150 mg format & has been optimized & validated for cell culture.

Synonym(s):

Primary Cell Culture Supplement

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Millipore, Upstate®

technique(s)

cell culture | mammalian: suitable

input

sample type neural stem cell(s)
sample type epithelial cells

shipped in

dry ice

storage temp.

-10 to -25°C

General description

ECGS is mitogenic for many types of cells under reduced or serum-free conditions, such as mammalian, avian and human endothelial cells, smooth muscle cells, keratinocytes, melanocytes and hybridomas; ECGS often fully substitutes for feeder layers in culture of fastidious cells.

Application

For use as a cell culture media supplement.
The Endothelial Cell Growth Supplement is designed for use as a cell culture media supplement. This cell growth supplement is available in a 150 mg format & has been optimized & validated for cell culture.

Physical form

150 mg, packaged in 10 bottles, each bottle containing 15 mg of bovine hypothalamic protein extract, rich in cell growth and attachment factors, lyophilized from sterile water containing trace amounts of NaCl and streptomycin sulfate.

Preparation Note

Lyophilized: Stable for 2 years at -20°C from date of shipment. Rehydrated: Stable for 3 months at -20°C.

Analysis Note

routinely evaluated in a 5 day growth assay of fetal bovine heart endothelial cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



S Strömblad et al.
The Journal of clinical investigation, 98(2), 426-433 (1996-07-15)
Induction of p53 activity in cells undergoing DNA synthesis represents a molecular conflict that can lead to apoptosis. During angiogenesis, proliferative endothelial cells become apoptotic in response to antagonists of integrin alphavbeta3 and this leads to the regression of angiogenic
H Hoshi et al.
In vitro cellular & developmental biology : journal of the Tissue Culture Association, 22(1), 51-56 (1986-01-01)
Endothelial cells from autopsy and biopsy specimens from a variety of adult human vascular tissue were harvested by collagenase treatment and gentle swabbing of the lumenal surface. Nutrient medium MCDB 107 containing a partially purified brain-derived growth factor (5 micrograms/ml)
N A Lanson et al.
Nucleic acids research, 28(15), 2882-2892 (2000-07-25)
To investigate molecular controls of cardiomyocyte proliferation, we utilized cardiomyocytes induced to proliferate indefinitely by SV40 large T antigen (T-ag). In the T-ag-immortalized AT-1, AT-2 and HL-1 cardiomyocytes, normal cellular proteins associating with T-ag and p53 were identified, isolated and



Global Trade Item Number

SKUGTIN
02-10204053252513060