Skip to Content
Merck
All Photos(1)

Key Documents

QGARD00R1

Milli-Q

Q-Gard® Purification Cartridge

For Milli-Q® Element (2001-2009) / Academic / Biocel / Element / Gradient / Synthesis systems connected to Elix® / RO / distilled water feed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Q-Gard®

resistivity

>18.0 MΩ-cm (at 25 °C)

technique(s)

ion chromatography: suitable

total organic carbon (TOC) residue

<10 ppb

capacity

77 g ion exchange capacity (NaCl)(Dynamic Capacity is more than 50 g NaCl)

compatibility

for use with Milli-Q® Academic
for use with Milli-Q® Biocel
for use with Milli-Q® Element
for use with Milli-Q® Gradient
for use with Milli-Q® Synthesis

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Q-Gard® 1 Purification Cartridge is designed to remove ionic and organic contaminants down to trace levels when fed with Elix® / RO / distilled water.
The cartridge is constructed with clean and high-purity materials to minimize leaching of extractable contaminants.
It contains a microfilter for particulate removal, activated carbon for organics adsorption, and mixed-bed ion-exchange resin for ion removal.
The Q-Gard® 1 Purification Cartridge can be used with Milli-Q® Academic / Biocel / Element / Gradient / Synthesis water purification systems.

Application

Removal of ionic and organic water contaminants.

Features and Benefits

  • Q-Gard® 1 Purification Cartridge is tailored to the feed-water source to optimize the performance of downstream purification media to ensure consistent results.
  • Genuine water purification consumable for replacement of the Q-Gard® QGARD00R1 cartridge filter.
  • Purification method involves ion-exchange, activated carbon, and depth filtration.
  • Certificate of Quality available online.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service