Skip to Content
Merck

Skip To

MPGP002A1

Millipak® Filter

0.22 μm membrane filter to remove particulates and bacteria at the point of dispense of Milli-Q® IQ/IX/EQ 7 water systems.

Millipak® Filter
2 of 5 reviewers received a sample product or took part in a promotion

Synonym(s):

Millipak® 0.22 μm filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104212
NACRES:
NB.24
eCl@ss:
33050190
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyethersulfone filter

Quality Segment

packaging

pkg of 1 unit

manufacturer/tradename

Millipak®

pore size

0.22 μm pore size

compatibility

for use with Milli-Q® EQ 7000, for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7000, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015

shipped in

ambient

General description

The Millipak® 0.22μm hydrophilic membrane filter is intended to remove particulates and bacteria from pure and ultrapure water at the point of dispense of the Milli-Q® IQ 7000/7003/05/10/15, Milli-Q® IX 7003/05/10/15 and Milli-Q® EQ 7000/08/16 water purification systems.
The filter is designed with a 0.22μm PES (polyethersulfone) asymmetric membrane filter heat-sealed on a styrene-acrylonitrile housing that serves to minimize organic and inorganic extractable release.

Application

Pure and Ultrapure water produced by a Millipak® 0.22 μm filter is suitable for many different laboratory applications, including microplastics research,[1] nanoparticle studies, food analyses and HPLC analyses.[2]

Features and Benefits

  • Each Millipak® 0.22μm filter is individually tested, hermetically sealed and delivered with a Certificate of Quality.
  • Validated to remove bacteria from water (<0.01cfu/mL when installed and used in a laminar flow hood).
  • Carefree maintenance: Easily installed and replaced.
  • Redesigned bottom-tip for a secured installation of the protection bell.
  • Validated to remove particulates from water (no particles with size >0.22 μm)
  • e-Sure tag for RFID connection with E-POD® or Q-POD® dispenser enables full traceability (data management) and automatic consumable status monitoring on the POD’s touchscreen interface.

Other Notes

Directions For Use:

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: General laboratory analysis
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 ?m sterilizing-grade membranes. Refer to the system equipment user guide section "Using the System" (available on USB key in the system box).
  • Storage Statement: Store in a dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

E-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MPGPG02A1PRPK000A1TANKVH1A1
description

0.22 μm membrane filter to remove particulates and bacteria at the point of dispense of Milli-Q® IQ/IX/EQ 7 water systems.

description

Removes particulates and bacteria at the point of dispense of Milli-Q® IQ/IX/EQ 7 water systems. Gamma-irradiated for additional bacterial control.

description

Additional pretreatment pack. Protects and maintains good performance of the reverse osmosis (RO) membrane. For Milli-Q® IQ 7003/05/10/15, Milli-Q® IX 7003/05/10/15 and Milli-Q® EQ 7008/16 systems.

description

Protects stored pure water from airborne contaminants. For Milli-Q® IQ/IX/EQ storage tanks. For High-Flow applications.

compatibility

for use with Milli-Q® EQ 7000, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7010, for use with Milli-Q® IX 7003, for use with Milli-Q® IQ 7000, for use with Milli-Q® IQ 7015, for use with Milli-Q® IX 7010, for use with Milli-Q® EQ 7008, for use with Milli-Q® IQ 7005

compatibility

for use with Milli-Q® EQ 7000, for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7000, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015

compatibility

for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015

compatibility

for use with Milli-Q® EQ 7008, for use with Milli-Q® EQ 7016, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.5 μm pore size (High capacity filter CRW05 type)

pore size

0.45 μm pore size

manufacturer/tradename

Millipak®

manufacturer/tradename

Millipak®

manufacturer/tradename

-

manufacturer/tradename

-

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 unit

material

polyethersulfone filter

material

polyethersulfone filter

material

polyethylene housing, polypropylene housing

material

-

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient


Still not finding the right product?

Explore all of our products under Millipak® Filter




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 11 Questions  
  1. Where can I find information on proper disposal and recycling of Milli-Q consumables (e.g. millipak filters)?

    1 answer
    1. Please follow your local regulations to dispose of filters as non-hazardous waste. If there are specific local regulations for electronic waste, we recommend that you dispose of RFID tags in accordance with these local regulations.

      Helpful?

  2. What are the storage conditions for this product?

    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.

      Helpful?

  3. How are consumables changed?

    1 answer
    1. To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.

      Helpful?

  4. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  5. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  6. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  7. Can consumables be opened and refilled?

    1 answer
    1. Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.

      Helpful?

  8. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  9. If I have a final filter that I use only occasionally (e.g., Biopak®) and have another filter (e.g. Millipak®) that I most of the time, can I keep the 'occasional-use' filter for use again at a later time?

    1 answer
    1. It is not possible to re-use a filter that has been used and then removed because air intake during replacement creates an airlock and blocks water passing through the filter. If you regularly need two different filters, we recommend using two POD dispensers.

      Helpful?

  10. What is the consumables delivery time?

    1 answer
    1. Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.

      Helpful?

1–10 of 11 Questions  

Reviews

Active Filters

  1. Verbania
    • Review 1
    • Votes 7
    5 out of 5 stars.

    Prestazioni molto buone

    ottimo prodotto, rispetto alla qualità ottenuta della filtrazione

    Translate with Google

    Helpful?

  2. 4 Ratings-only reviews