Skip to Content
Merck
All Photos(1)

Key Documents

NA13

Sigma-Aldrich

Anti-Replication Protein A (Ab-1) Mouse mAb (RPA70-9)

liquid, clone RPA70-9, Calbiochem®

Synonym(s):

Anti-RP-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RPA70-9, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, yeast

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RPA1(6117)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with the NS1 myeloma cell line. Recognizes the ~70 kDa Replication Protein A.
Recognizes the ~70 kDa subunit of replication protein A in HeLa and u293 cells and colon carcinoma tissue.
This Anti-Replication Protein A (Ab-1) Mouse mAb (RPA70-9) is validated for use in Frozen Sections, Immunoblotting, IF, IP, Paraffin Sections for the detection of Replication Protein A (Ab-1).

Immunogen

Epitope: Within the p70 subunit of replication protein A
Human
replication protein A purified from U293 cells

Application


Frozen Sections (2.5 g/ml)
Immunoblotting (1-5 g/ml)
Immunofluorescence (2.5 g/ml)
Immunoprecipitation (1 g/reaction)
Paraffin Sections (2.5 g/ml, no pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.05 M sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Positive Control
HeLa or U293 cells or colon carcinoma tissue

Other Notes

Din, S., et al. 1990. Genes Dev.4, 968.
Brill, S.J. and Stillman, B., 1989. Nature342, 92.
Stillman, B., 1989. Annu. Rev. Cell. Biol.5, 197.
Tsurimoto, T. and Stillman, B., 1989. EMBO J.8, 3883.
Wobbe, C.R., et al. 1987. Proc. Natl. Acad. Sci. USA84, 1834.
Specific for the p70 subunit of replication protein A. This antibody can be used as a marker for proliferation. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Rodier et al.
Journal of cell science, 124(Pt 1), 68-81 (2010-12-02)
DNA damage can induce a tumor suppressive response termed cellular senescence. Damaged senescent cells permanently arrest growth, secrete inflammatory cytokines and other proteins and harbor persistent nuclear foci that contain DNA damage response (DDR) proteins. To understand how persistent damage
Ken-ichi Yoshioka et al.
Molecular cell, 22(4), 501-510 (2006-05-23)
S(N)1-type alkylating agents that produce cytotoxic O(6)-methyl-G (O(6)-meG) DNA adducts induce cell cycle arrest and apoptosis in a manner requiring the DNA mismatch repair (MMR) proteins MutSalpha and MutLalpha. Here, we show that checkpoint signaling in response to DNA methylation
S Buchsbaum et al.
Oncogene, 26(35), 5132-5144 (2007-02-22)
The mouse int6 gene is a frequent integration site of the mouse mammary tumor virus and INT6 silencing by RNA interference in HeLa cells causes an increased number of cells in the G2/M phases of the cell cycle, along with
Tom Stiff et al.
Human molecular genetics, 25(8), 1574-1587 (2016-02-26)
Mutations in ATR(ataxia telangiectasia and RAD3-related) cause Seckel syndrome (ATR-SS), a microcephalic primordial dwarfism disorder. Hitherto, the clinical manifestation of ATR deficiency has been attributed to its canonical role in DNA damage response signalling following replication fork stalling/collapse. Here, we
Yuliang Wu et al.
Biochemistry, 47(18), 5068-5077 (2008-04-16)
Alternate DNA structures other than double-stranded B-form DNA can potentially impede cellular processes such as transcription and replication. The DNA triplex helix and G4 tetraplex structures that form by Hoogsteen hydrogen bonding are two examples of alternate DNA structures that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service