Skip to Content
Merck
All Photos(2)

Key Documents

MABC203

Sigma-Aldrich

Anti-Yes-associated protein (YAP1) Antibody, clone 4D11.1

clone 4D11.1, from mouse

Synonym(s):

Yorkie homolog, Yes-associated protein, Yap65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D11.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... YAP1(10413)

General description

The YAP1 gene encodes a protein that binds to the SH3 (Src homology 3) domain of the Yes proto-oncogene product. As a transcriptional regulator, it can act as a coactivator and a corepressor, and serves as a critical downstream regulatory target in the Hippo signaling pathway. Additionally, it plays a critical role in regulating organ size and in tumor suppression by restricting proliferation and promoting apoptosis. The human YAP1 gene is well conserved among higher eukaryotes, but may not be conseved in yeast. Yes-associated protein expression is almost ubiquitous in adult human tissues, but is not detectable in peripheral blood leukocytes.

Immunogen

GST-tagged recombinant protein corresponding to human Yes-associated protein (YAP1).

Application

Anti-Yes-associated protein (YAP1) Antibody, clone 4D11.1 detects level of Yes-associated protein (YAP1) & has been published & validated for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Yes-associated protein (YAP1) in HeLa and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Yes-associated protein (YAP1) in 10 µg of HeLa cell lysate.

Target description

~65 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Hela cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service