MAB9012
Anti-Coronavirus Antibody, OC-43 strain, clone 541-8F
clone 541-8F, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
541-8F, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
shipped in
wet ice
Specificity
Coronavirus.
Immunogen
Epitope: OC-43 strain
Application
Anti-Coronavirus Antibody, OC-43 strain, clone 541-8F detects level of Coronavirus & has been published & validated for use in IF.
IFA. Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Format: Purified
Purified immunoglobulin. Supplied in PBS buffer pH 7.4. Contains 0.1% sodium azide and carrier protein.
Storage and Stability
Maintain at 2-8°C for up to 6 months from date of receipt.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of virology, 95(12) (2021-04-02)
The COVID-19 pandemic poses a serious global health threat. The rapid global spread of SARS-CoV-2 highlights an urgent need to develop effective therapeutics for blocking SARS-CoV-2 infection and spread. Stimulator of Interferon Genes (STING) is a chief element in host
Frontiers in microbiology, 13, 845269-845269 (2022-06-28)
The human coronavirus OC43 (HCoV-OC43) is one of the most common causes of common cold but can lead to fatal pneumonia in children and elderly. However, the available animal models of HCoV-OC43 did not show respiratory symptoms that are insufficient
Frontiers in cellular and infection microbiology, 12, 850744-850744 (2022-05-14)
The endemic and pandemic caused by respiratory virus infection are a major cause of mortality and morbidity globally. Thus, broadly effective antiviral drugs are needed to treat respiratory viral diseases. Small extracellular vesicles derived from human umbilical cord mesenchymal stem
Scientific reports, 12(1), 14438-14438 (2022-08-25)
The lack of therapeutic options to fight Covid-19 has contributed to the current global pandemic. Despite the emergence of effective vaccines, development of broad-spectrum antiviral treatment remains a significant challenge, in which antimicrobial photodynamic therapy (aPDT) may play a role
Phytomedicine : international journal of phytotherapy and phytopharmacology, 93, 153808-153808 (2021-11-10)
Qingwenjiere Mixture (QJM) is a traditional Chinese medicine (TCM) that has been shown to have remarkable clinical efficacy against COVID-19. However, little is known about the antiviral and anti-inflammatory activities of QJM against a wider range of human coronavirus (HCoV)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service