Skip to Content
Merck
All Photos(2)

Key Documents

N2912

Sigma-Aldrich

Anti-Neurofilament 160/200 antibody, Mouse monoclonal

~2 mg/mL, clone RMdO20, purified from hybridoma cell culture

Synonym(s):

Anti-NEF3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

RMdO20, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
microarray: suitable
western blot: 1-2 μg/mL using extracts of rat brain S1 fraction.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Neurofilaments are 10 nm in diameter and belong to intermediate filament protein family. Neurofilaments are expressed mainly in cells or tissues of neuronal origin and are crucial for proper radial growth of axons. Monoclonal anti-neurofilament 160/200 antibody can be used in ELISA and immunoblotting. It can also be used in microarray. Mouse anti-neurofilament 160/200 antibody reacts specifically with the non-phosphorylated form of NF-M and NF-H (NF-M/H, approx.160 and 200 kDa) in mouse, rat and human.

Immunogen

purified mid-size rat neurofilament (NF-M) subunit.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)
Monoclonal anti-neurofilament 160/200 antibody can be used in immunocytochemistry (diluted 1: 100) and western blotting (diluted 1: 1,000) . It can also be used in immunohistochemistry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kim Braeckman et al.
NeuroImage. Clinical, 21, 101669-101669 (2019-01-20)
Diffusion magnetic resonance imaging biomarkers can provide quantifiable information of the brain tissue after a mild traumatic brain injury (mTBI). However, the commonly applied diffusion tensor imaging (DTI) model is not very specific to changes in the underlying cellular structures.
L Yao et al.
Gene therapy, 20(12), 1149-1157 (2013-07-26)
Functionalized biomaterial scaffolds targeted at improving axonal regeneration by enhancing guided axonal growth provide a promising approach for the repair of spinal cord injury. Collagen neural conduits provide structural guidance for neural tissue regeneration, and in this study it is
M K Lee et al.
Current opinion in cell biology, 6(1), 34-40 (1994-02-01)
Neurofilaments make up the major intermediate filament system in mature neurons. Recent studies demonstrate that neurofilaments in vivo are obligate heteropolymers and are required for proper radial growth of axons. Furthermore, forced over-expression of neurofilament subunits in transgenic mice shows
Yaxian Wang et al.
International journal of molecular sciences, 18(2) (2017-01-31)
Peripheral nerve injury triggers the dysregulation of a large number of genes at multiple sites, including neurons, peripheral nerve stump, and the target organ. Housekeeping genes were frequently used as reference genes to normalize the expression values of target genes.
Mariangela Iovino et al.
Brain : a journal of neurology, 138(Pt 11), 3345-3359 (2015-07-30)
Tauopathies, such as Alzheimer's disease, some cases of frontotemporal dementia, corticobasal degeneration and progressive supranuclear palsy, are characterized by aggregates of the microtubule-associated protein tau, which are linked to neuronal death and disease development and can be caused by mutations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service