Skip to Content
Merck
All Photos(2)

Key Documents

MAB1952Z

Sigma-Aldrich

Anti-Integrin α3 Antibody, clone P1B5, azide free

clone P1B5, Chemicon®, from mouse

Synonym(s):

CD49c, MAB1952

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1B5, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

General description

Integrins are a family of dimeric, transmembrane proteins that mediate cell-cell and extracellular matrix adhesion. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. The integrin family is composed of at least 15 alpha and 8 beta subunits that may form over twenty different alpha-beta non-covalently bound dimeric combinations on the cell surface. The alpha subunits all have some homology to each other, as do the beta subunits. Both of the subunits contribute to the binding of the ligand. Integrin alpha subunits contain seven weak sequence repeats in the N-terminal region, which may be important in ligand binding, and have been predicted to fold cooperatively into a single beta-propeller domain with seven beta-sheets. The alpha-3 subunit (CD49c) is highly concentrated in epithelial cells where it strongly adheres to Laminin-5 and Laminin-5 induced rapid adhesion can be blocked by antibodies against the alpha-3 integrin subunit. The alpha-3 subunit exists in two different splice variants, denoted as "A" and "B". The only difference that results from this differential splicing is a total change in the cytoplasmic domain, while the extracellular domain stays the same. Knock-out mice lacking this subunit show prenatal lethality and abnormalities in the kidneys.

Specificity

Reacts with Human alpha3 integrin.

Application

Anti-Integrin α3 Antibody, clone P1B5, azide free detects level of Integrin α3 & has been published & validated for use in IC, IH(P).
Research Category
Cell Structure
Research Sub Category
Integrins
Suitable for use in attachment inhibition assays using fibroblasts, most epithelial cells, activated lymphocytes on laminin, collagen and fibronectin.

Immunohistochemistry on human tonsil

fresh frozen tissue

paraffin embedded tissue; protease digestion required:

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with no preservatives.
Protein A purified

Storage and Stability

Maintain at 2–8°C in undiluted aliquots for up to 6 months after date of receipt.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Analysis Note

Control
Human foetal foreskin fibroblasts (HFFF), neuroblastomas

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-Yi Ke et al.
Cancer science, 111(10), 3478-3492 (2020-07-11)
Ovarian cancer has a high recurrence rate after platinum-based chemotherapy. To improve the treatment of ovarian cancer and identify ovarian cancer-specific antibodies, we immunized mice with the human ovarian carcinoma cell line, SKOV-3, and generated hybridoma clones. Several rounds of
Anna Valeria Samarelli et al.
The Journal of biological chemistry, 289(28), 19466-19476 (2014-05-27)
The synaptic protein Neuroligin 1 (NLGN1), a cell adhesion molecule, is critical for the formation and consolidation of synaptic connectivity and is involved in vascular development. The mechanism through which NLGN1 acts, especially in vascular cells, is unknown. Here, we
J Speer et al.
European cells & materials, 41, 793-810 (2021-06-24)
With aging and pathology, cells of the nucleus pulposus (NP) de-differentiate towards a fibroblast-like phenotype, a change that contributes to degeneration of the intervertebral disc (IVD). Laminin isoforms are a component of the NP extracellular matrix during development but largely
Richard A Que et al.
Journal of biomedical materials research. Part A, 106(5), 1363-1372 (2018-01-18)
Adhesion to the microenvironment profoundly affects stem cell functions, including proliferation and differentiation, and understanding the interaction of stem cells with the microenvironment is important for controlling their behavior. In this study, we investigated the effects of the integrin binding
James Kenney et al.
PloS one, 16(7), e0254714-e0254714 (2021-07-17)
Integrin receptors for the extracellular matrix play critical roles at all stages of carcinogenesis, including tumor growth, tumor progression and metastasis. The laminin-binding integrin α3β1 is expressed in all epithelial tissues where it has important roles in cell survival, migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service