Skip to Content
Merck

AB1936

Anti-Integrin α2 Antibody

serum, Chemicon®

Synonym(s):

CD49b

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IF, IHC, IP, RIA, WB
Citations:
50
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

chicken, rat, hamster, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, radioimmunoassay: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA2(3673)

General description

160kDa

Immunogen

Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human alpha2 integrin subunit {KYEKMTKNPDEODETTELSS}

Application

Anti-Integrin α2 Antibody detects level of Integrin α2 & has been published & validated for use in ELISA, IF, IH, IP, RIA & WB.
Immunofluorescence: 1:500 (suggested for use on acetone-fixed tissue only)

ELISA/RIA: 1:1000

Western blot: 1:1000 The antibody has been used to report alpha2 expression in HUVECs during tubule-like structure formation in collagen (Deroanne, 1996). Recognizes ~160kDa alpha 2 subunit

Immunoprecipitation: 5 μL precipitates alpha2/beta1 from 5 x 106 cells.

Immunohistochemistry: 1:500

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Biochem/physiol Actions

Human alpha2 integrin subunit. Specificity determined by immunoprecipitation from a detergent extract of 35S methionine labeled endothelial cells. The epitope recognized is on the cytoplasmic domain of the protein and is thus intracellular. Shows no cross-reactivity to alpha1, alpha3, alpha4, alpha5, alpha6, or alphaV subunits.

Physical form

Neat rabbit antiSerum. Liquid with 0.05% sodium azide
Unpurified

Preparation Note

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin (basement membrane), High grade ovarian tumors

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library