Skip to Content
Merck
All Photos(3)

Key Documents

F2518

Sigma-Aldrich

Human Fibronectin

from human foreskin fibroblasts, powder, suitable for cell culture

Synonym(s):

Fibronectin

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Fibronectin human foreskin fibroblasts, lyophilized powder, suitable for cell culture

biological source

human foreskin fibroblasts

Quality Level

product line

BioReagent

form

lyophilized powder

mol wt

~550 kDa

packaging

pkg of 0.5 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑5 μg/cm2

impurities

Human Source, none detected (HCV)
Human Source, none detected (HIV)
Human Source, none detected (HTLV)
Human Source, none detected (HbSAg)
Microbial Contamination, passes test

solubility

H2O: 1 mg/mL at 37 °C (Store reconstituted solution in working aliquots at -20°C or lower.)

NCBI accession no.

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin 1 (FN1) is a glycoprotein of the extracellular matrix that is coded by FN1 gene. It is expressed in the plasma and at the cell surface. It is mapped to human chromosome 2q35.[1] Fibronectin is mostly synthesized by fibroblasts and endothelial cells.[2]It is an adhesive glycoprotein,[3] predominant in extracellular matrix (ECM) and is widely distributed in tissue and blood. The fibronectin structure consists of blocks of repeating peptide sequences that are homologous and form functional units arranged linearly ontwo subunit arms.[4]

Application

Epithelial cells, mesenchymal cells, neuronal cells, fibroblasts, neural crest cells, endothelial cells
Fibronectin human foreskin fibroblasts has been used in extracellular matrix binding assay[5][6] and probiglycan and cleavage assays.[7] It has also been used in ectoderm (neuronal) differentiation[8] and in the preparation of a semi-solid growth medium.[9]

Biochem/physiol Actions

Fibronectin 1 (FN1) participates in cell adhesion, growth, migration, wound healing, blood coagulation and metastasis.[1] Mutations in FN1 results in glomerulopathy. It plays an important role in cell attachment and spreading, control of cell cytoskeleton, morphology and differentiation. FN1 is also involved in extracellular matrix formation, hemostasis and thrombosis.[10] Fibronectin production by normal human foreskin fibroblasts has been shown to improve two-fold after glucocorticoid treatment.[2]

Features and Benefits

  • Fibronectin is isolated from human foreskin fibroblasts maintained in serum-free medium.
  • It can be used as a cell attachment factor for various cell types, including epithelial, mesenchymal, neuronal, neural crest, endothelial cells, and fibroblasts.
  • The starting material is tested and found negative for HIV and HBsAg.
  • The product is soluble in sterile water, yielding a clear, colorless solution.
  • It can be stored at -20°C for at least three years.

Caution

Vortexing, excessive agitation, repeated freezing and thawing of reconstituted fibronectin are not recommended.

Physical form

Lyophilized in CAPS buffered saline

Preparation Note

dialyzed against chloroform

Reconstitution

Store reconstituted solution in working aliquots at -20°C or lower.

Analysis Note

Protein homogeneity is evaluated by immunoelectrophoresis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibronectin
Fabrizio M and Angela S
Encyclopedia of Immunology, 909-913 (1998)
Regulation of matrix Accumulation (2012)
Ail binding to fibronectin facilitates Yersinia pestis binding to host cells and Yop delivery.
Tsang T M, et al.
Infection and Immunity, 78(8), 3358-3368 (2010)
Artificial biomaterials for urological tissue engineering
Farhat WA and Geutjes, PJ
Journal of Separation Science (2009)
Fibronectin: a brief overview of its structure, function, and physiology
Proctor RA
Reviews of Infectious Diseases, 317?321-317?321 (1987)

Articles

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

See All

Protocols

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service